Skip to Content
Merck
All Photos(1)

Key Documents

342041

Sigma-Aldrich

Perfluoroheptanoic acid

≥97.0%

Synonym(s):

Tridecafluoroheptanoic acid, Perfluoroheptanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3(CF2)5CO2H
CAS Number:
Molecular Weight:
364.06
Beilstein:
1808210
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0%

form

solid

refractive index

n20/D 1.306 (lit.)

bp

175 °C/742 mmHg (lit.)

mp

30 °C (lit.)

density

1.792 g/mL at 25 °C (lit.)

SMILES string

OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C7HF13O2/c8-2(9,1(21)22)3(10,11)4(12,13)5(14,15)6(16,17)7(18,19)20/h(H,21,22)

InChI key

ZWBAMYVPMDSJGQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Perfluoroheptanoic acid can be used as a reactant:
  • For the fluoroalkylation of C-H bonds of aromatic compounds using Ni/SiO2 nanocatalyst.
  • To synthesize biphenylyl benzoates via Mitsunobu reaction.
  • To synthesize N-perfluoracyl β-thioamides from β-thio amines in the presence of methyl chloroformate.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B - STOT RE 1

Target Organs

Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bart B L Groen et al.
The Journal of clinical endocrinology and metabolism, 101(11), 3978-3988 (2016-11-05)
Skeletal muscle protein synthesis is highly responsive to food intake. It has been suggested that the postprandial increase in circulating insulin modulates the muscle protein synthetic response to feeding. The objective of the study was to investigate whether a greater
N Kudo et al.
Chemico-biological interactions, 134(2), 203-216 (2001-04-20)
Elimination in urine and feces was compared between four perfluorinated fatty acids (PFCAs) with different carbon chain length. In male rats, perfluoroheptanoic acid (PFHA) was rapidly eliminated in urine with the proportion of 92% of the dose being eliminated within
Synthesis of new fluorous modular chiral ligand derivatives from amino alcohols and application in enantioselective carbon-carbon bond-forming alkylation reactions
Sehnem J, et al.
Tetrahedron Asymmetry, 21, 997-1003 (2010)
Mikhail N Khrizanforov et al.
Dalton transactions (Cambridge, England : 2003), 45(30), 11976-11982 (2016-07-08)
We have developed Ni(III)-doped silica nanoparticles ([(bpy)xNi(III)]@SiO2) as a recyclable, low-leaching, and efficient oxidative functionalization nanocatalyst for aromatic C-H bonds. The catalyst is obtained by doping the complex [(bpy)3Ni(II)] on silica nanoparticles along with its subsequent electrooxidation to [(bpy)xNi(III)] without
Wenchao Han et al.
Environment international, 116, 206-213 (2018-04-27)
Perfluoroalkyl and polyfluoroalkyl substances (PFASs) have been widely detected in different populations. However, limited data is available about exposure among family members. To investigate the PFASs levels in parents and their newborns and to understand their correlation and health implications

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service