Skip to Content
Merck
All Photos(3)

Key Documents

380008

Sigma-Aldrich

Ammonium bromide

99.999% trace metals basis

Synonym(s):

Azanium bromide, Hydrobromic acid monoammoniate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4Br
CAS Number:
Molecular Weight:
97.94
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38050407
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

Quality Level

vapor pressure

1 mmHg ( 198.3 °C)

Assay

99.999% trace metals basis

form

powder

impurities

≤15.0 ppm Trace Metal Analysis

mp

452 °C (lit.)

density

2.43 g/mL at 25 °C (lit.)

SMILES string

N.Br[H]

InChI

1S/BrH.H3N/h1H;1H3

InChI key

SWLVFNYSXGMGBS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Eye Irrit. 2 - Lact. - Repr. 1B - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catalytic synthesis of nonracemic azaproline derivatives by cyclization of β-alkynyl hydrazines under kinetic resolution conditions.
Pradip Maity et al.
Angewandte Chemie (International ed. in English), 50(36), 8338-8341 (2011-07-19)
Erhong Duan et al.
Journal of hazardous materials, 194, 48-52 (2011-08-27)
The solubility of SO(2) in a binary mixture of water and caprolactam tetrabutyl ammonium bromide ionic liquid (CPL-TBAB IL) was investigated. Though the ionic liquid and water were fully miscible, a phase separation occurred when SO(2) was introduced into the
Fredrik Hallberg et al.
Journal of magnetic resonance (San Diego, Calif. : 1997), 192(1), 69-77 (2008-02-22)
Although simple as a concept, electrophoretic NMR (eNMR) has so far failed to find wider application. Problems encountered are mainly due to disturbing and partly irreproducible convection-like bulk flow effects from both electro-osmosis and thermal convection. Additionally, bubble formation at
Zhen Chen et al.
Physical review. E, Statistical, nonlinear, and soft matter physics, 84(1 Pt 1), 011401-011401 (2011-08-27)
The phase behavior of an inverse microemulsion composed of 4-cyano-4-n-pentylbiphenyl (5CB), didodecyl dimethyl ammonium bromide, and water was intensively studied recently, but controversy still remains on the existence of a "transparent nematic" (TN) phase. Previously, we studied the temperature-dependent dielectric
Dong Xie et al.
Dental materials : official publication of the Academy of Dental Materials, 27(5), 487-496 (2011-03-11)
The objective of this study was to use the newly synthesized poly(quaternary ammonium salt) (PQAS)-containing polyacid to formulate the light-curable glass-ionomer cements and study the effect of the PQAS on the compressive strength and antibacterial activity of the formed cements.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service