Skip to Content
Merck
All Photos(3)

Key Documents

372862

Sigma-Aldrich

Sodium perborate monohydrate

powder, 20-100 mesh

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BNaO3 · H2O
CAS Number:
Molecular Weight:
99.81
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

form

powder

concentration

10.3-11.4% B (by NaOH, titration)

particle size

20-100 mesh

SMILES string

[Na+].[H]O[H].[O-]B1OO1

InChI

1S/BO3.Na.H2O/c2-1-3-4-1;;/h;;1H2/q-1;+1;

InChI key

VBLJKTBPWITVTE-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Ox. Sol. 3 - Repr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ingrid Machado de Andrade et al.
Gerodontology, 28(4), 264-270 (2010-06-04)
To evaluate the antimicrobial action of effervescent tablets and ultrasound on Candida spp. and mutans streptococci from denture biofilm. It is not uncommon for edentulous patients to be elderly and find it difficult to brush their dentures. Hence, auxiliary methods
Eduardo Buozi Moffa et al.
Journal of dentistry, 39 Suppl 3, e65-e71 (2011-11-03)
This randomised clinical study evaluated the effect of chemical disinfection with sodium perborate or chlorhexidine on the colour stability of a hard chairside reline resin during six months. Hard chairside reline resin (Tokuyama Rebase Fast II) was used to reline
Milton Carlos Kuga et al.
Dental traumatology : official publication of International Association for Dental Traumatology, 28(3), 238-242 (2011-11-22)
To compare the fracture resistance of bovine teeth after intracoronal bleaching with sodium percarbonate (SPC) or sodium perborate (SP) mixed with water or 20% hydrogen peroxide (HP). Fifty extracted bovine teeth were divided into four experimental groups (G1-G4) and one
Divya S Sharma et al.
The Journal of clinical pediatric dentistry, 35(4), 349-352 (2011-11-04)
The odd attracts society in odd manner, as is the case when a patient with discolored tooth smiles. Because of that, pediatric patients have psychological impact. Trauma and pulpal necrosis are the most common causes for discoloration of teeth. If
Daniela Nair Borges Felipucci et al.
Brazilian dental journal, 22(5), 392-397 (2011-10-21)
Removable partial dentures (RPD) demand specific hygienic cleaning and the combination of brushing with immersion in chemical solutions has been the most recommended method for control of biofilm. However, the effect of the cleansers on metallic components has not been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service