Skip to Content
Merck
All Photos(4)

Key Documents

D3412

Sigma-Aldrich

Sodium 1-decanesulfonate

~98%

Synonym(s):

1-Decanesulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)8CH2SO3Na
CAS Number:
Molecular Weight:
244.33
Beilstein:
3918920
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

description

anionic

Assay

~98%

form

powder

mol wt

244.33 g/mol

technique(s)

LC/MS: suitable

mp

>300 °C (lit.)

SMILES string

[Na+].CCCCCCCCCCS([O-])(=O)=O

InChI

1S/C10H22O3S.Na/c1-2-3-4-5-6-7-8-9-10-14(11,12)13;/h2-10H2,1H3,(H,11,12,13);/q;+1/p-1

InChI key

AIMUHNZKNFEZSN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium 1-decanesulfonate is an anionic surfactant that is used for the analysis of organic small molecule compounds by HPLC and by ion-pair liquid chromatography. The anionic sulfonate counterion permits the separation and resolution of positively charged analytes. The critical micelle concentration (CMC) of sodium 1-decanesulfonate is assessed using capillary electrophoresis. Some studies report the interactions between hydroxypropylmethylcellulose and several ionic surfactants, like sodium 1-decanesulfonate.

Application

Ion-associating reagent for HPLC, including analyses of peptides and proteins.
Sodium 1-decanesulfonate has been used in a study to assess the mechanism of Zn2+ ions electroreduction. It has also been used in a study to investigate the retention mechanism of guanidine compounds on a porous graphitic carbon.

Features and Benefits

  • Suitable for high-performance liquid chromatography (HPLC) and ion-pair liquid chromatography
  • Allows the separation and resolution of positively charged analytes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

J J Berzas Nevado et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 769(2), 261-268 (2002-05-09)
A micellar electrokinetic capillary chromatography (MEKC) for determining fluoxetine and its metabolite (norfluoxetine) is proposed. Optimal conditions for the quantitative separation were investigated. A background electrolyte solution consisting of 5 mM phosphate buffer adjusted to pH 12.3 and 40 mM
Y Inamoto et al.
Journal of chromatography. B, Biomedical sciences and applications, 707(1-2), 111-120 (1998-06-05)
The retention mechanism of guanidino compounds on a porous graphitic carbon seemed to be mainly hydrophobic interaction, according to the retention factors in buffer solutions and the results of an analysis by computational chemical calculation using molecular mechanics (MM2). The
Influence of sodium 1-decanesulfonate on the enthalpy of activation of two-step Zn2+ ions electroreduction
Nieszporek
Journal of Electroanalytical Chemistry (Lausanne Switzerland), 662, 8-8 (2011)
W A Moats
Journal of agricultural and food chemistry, 48(6), 2244-2248 (2000-07-11)
A simplified procedure was developed for determination of tetracycline antibiotics in tissues which improved stability of these compounds in sample extracts and eliminated the need for troublesome cleanup procedures. Tissues were homogenized in water. Acetonitrile (16 mL) and then 1
N M Kovalchuk et al.
Journal of colloid and interface science, 459, 250-256 (2015-08-25)
Mixed solutions of cationic and anionic surfactants show considerable synergism in their wetting behaviour, but their spreading is affected considerably by the phase separation processes. The valuable information about wetting properties of synergetic mixtures can be obtained by using mixtures

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service