Skip to Content
Merck
All Photos(4)

Key Documents

243655

Sigma-Aldrich

Sodium molybdate

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2MoO4
CAS Number:
Molecular Weight:
205.92
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55
Assay:
≥98%
form:
granular

Quality Level

Assay

≥98%

form

granular

reaction suitability

reagent type: catalyst
core: molybdenum

mp

687 °C (lit.)

density

3.78 g/mL at 25 °C (lit.)

SMILES string

[Na+].[Na+].[O-][Mo]([O-])(=O)=O

InChI

1S/Mo.2Na.4O/q;2*+1;;;2*-1

InChI key

TVXXNOYZHKPKGW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium molybdate has corrosion resistant properties. On heating, it undergoes degradation and releases Na2O fumes.

Application

Sodium molybdate (Na2MO4) may be used as an electrolyte additive, for use in electrochemical capacitors. Addition of Na2MO4 as an electrolyte additive may result in enhanced capacitance, corrosion prevention and stabilized performance.
It may also be used as a catalyst during the manufacture of paints and dyes.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Q Abbas et al.
Faraday discussions, 172, 199-214 (2014-11-27)
Sodium molybdate (Na2MoO4) has been used as an additive to 1 mol L(-1) lithium sulfate electrolyte for electrochemical capacitors based on activated carbon (AC) electrodes, in order to reduce the corrosion of stainless steel current collectors. We demonstrate that the
Ash M and Ash I.
Handbook of Green Chemicals, 878-878 (2004)
Jason Davenport et al.
Journal of natural products, 74(5), 1085-1092 (2011-04-14)
A high-throughput screening of natural product libraries identified (-)-gambogic acid (1), a component of the exudate of Garcinia harburyi, as a potential Hsp90 inhibitor, in addition to the known Hsp90 inhibitor celastrol (2). Subsequent testing established that 1 inhibited cell
Bo-Geon Yun et al.
Biochemistry, 43(25), 8217-8229 (2004-06-24)
Hsp90 functions to facilitate the folding of newly synthesized and denatured proteins. Hsp90 function is modulated through its interactions with cochaperones and the binding and hydrolysis of ATP. Recently, novobiocin has been shown to bind to a second nucleotide binding
Vaddypally Shivaiah et al.
Inorganic chemistry, 44(24), 8846-8854 (2005-11-22)
Two Anderson-type heteropolyanion-supported copper phenanthroline complexes, [Al(OH)6Mo6O18[Cu(phen)(H2O)2]2]1+ (1c) and [Al(OH)6Mo6O18[Cu(phen)(H2O)Cl]2]1- (1a) complement their charges in one of the title compounds [Al(OH)6Mo6O18[Cu(phen)(H2O)2]2][Al(OH)6Mo6O18[Cu(phen)(H2O)Cl]2].5H2O [1c][1a].5 H2O 1. Similar charge complementarity exists in the chromium analogue, [Cr(OH)6Mo6O18[Cu(phen)(H2O)2]2][Cr(OH)6Mo6O18[Cu(phen)(H2O)Cl]2].5 H2O [2c][2a].5 H2O 2. The chloride coordination

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service