Skip to Content
Merck
All Photos(2)

Key Documents

369756

Sigma-Aldrich

2-Phenyl-2-oxazoline

99%

Synonym(s):

2-Phenyl-1,3-oxazoline, 2-Phenyl-4,5-dihydro-1,3-oxazole, 2-Phenyl-4,5-dihydrooxazole, 4,5-Dihydro-2-phenyloxazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9NO
CAS Number:
Molecular Weight:
147.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.567 (lit.)

bp

75 °C/0.3 mmHg (lit.)

mp

12 °C (lit.)

density

1.118 g/mL at 25 °C (lit.)

functional group

ether
phenyl

SMILES string

C1CN=C(O1)c2ccccc2

InChI

1S/C9H9NO/c1-2-4-8(5-3-1)9-10-6-7-11-9/h1-5H,6-7H2

InChI key

ZXTHWIZHGLNEPG-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ignacio Del Río et al.
Acta crystallographica. Section E, Structure reports online, 65(Pt 1), m103-m104 (2008-01-01)
The title material, [Zn(C(5)H(7)O(2))(2)(C(9)H(9)NO)], was synthesized by the treatment of bis-(acetyl-acetonato)zinc(II) monohydrate with 2-phenyl-2-oxazoline. The Zn atom is coordinated by two chelating acetyl-acetonate groups and one oxazoline ligand in the apical position of a slightly distorted square-pyramidal metal-ligand geometry.
Robert A Gossage et al.
Dalton transactions (Cambridge, England : 2003), (23)(23), 3115-3122 (2008-06-04)
The treatment of cold ( approximately 3 degrees C) methanolic solutions of Li(2)PdCl(4) with two equivalents of 2-phenyl-2-oxazoline (Phox) results in the isolation of [PdCl(2)(Phox)(2)] (3). This complex undergoes remarkably slow isomerisation (CHCl(3)-d) at room temperature to a corresponding thermodynamic
One-pot synthesis of 2-phenyl-2-oxazoline-containing quasi-diblock copoly (2-oxazoline) s under microwave irradiation.
Hoogenboom R, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 45(3), 416-422 (2007)
Microwave Accelerated Polymerization of 2-Phenyl-2-oxazoline.
Sinnwell S and Ritter H.
Macromolecular Rapid Communications, 26(3), 160-163 (2005)
Andrew M Kelly et al.
Macromolecular rapid communications, 32(22), 1815-1819 (2011-09-21)
A 32-membered library of poly(2-oxazoline)-based hydrogels of the composition pEtOx(m) -pPhOx(n) -pPBO(q) (m/n = 150/0, 100/50, 50/100, and 0/150; q = 1.5-30) was prepared from 2-ethyl- (EtOx), 2-phenyl-2-oxazoline (PhOx), and phenylene-1,3-bis-(2-oxazoline) (PBO). The polymerizations were performed from ground monomer mixtures

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service