Skip to Content
Merck
All Photos(1)

Documents

129445

Sigma-Aldrich

Indole-3-carboxaldehyde

97%

Synonym(s):

β-Indolylaldehyde, 3-Formylindole, 3-Indolylformaldehyde, Indole-3-carbaldehyde, NSC 10118

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7NO
CAS Number:
Molecular Weight:
145.16
Beilstein:
114117
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

193-198 °C (lit.)

SMILES string

O=Cc1c[nH]c2ccccc12

InChI

1S/C9H7NO/c11-6-7-5-10-9-4-2-1-3-8(7)9/h1-6,10H

InChI key

OLNJUISKUQQNIM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Indole-3-carboxaldehyde can undergo Schiff bases condensation to form multifunctional silica nano-vehicles and magnetic nanoparticles.

Application

Indole-3-carboxaldehyde was used to prepare analogs of the indole phytoalexin cyclobrassinin with NR1R2 group. It was also used as the starting material for the synthesis of higher order indoles including isoindolo[2,1-a]indoles, aplysinopsins, and 4-substituted-tetrahydrobenz[cd]indoles.
Reactant for preparation of:
  • Analgesic agents
  • Hypoglycemic agents
  • Tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators
  • Antibacterial and antifungal agents
  • Antiamoebic and cytotoxic agents
  • Inhibitors of the Dengue virus protease with antiviral activity in cell-culture
  • Curcumin analogues as possible anti-proliferative & anti-inflammatory agents
  • Inhibitors of Bcl-2 family proteins
  • Inhibitors of the C-terminal domain of RNA Polymerase II as antitumor agents
  • Inhibitors of TNF-α and IL-6 with anti-tubercular activity

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Indian J. Chem. B, 33, 4-4 (1994)
Heterocycles, 38, 1479-1479 (1994)
Mariana Budovská et al.
Bioorganic & medicinal chemistry, 21(21), 6623-6633 (2013-09-10)
An effective synthesis of analogs of the indole phytoalexin cyclobrassinin with NR1R2 group instead of SCH3 was developed starting from indole-3-carboxaldehyde. The target compounds were prepared by spirocyclization of 1-Boc-thioureas with the formation of isolable spiroindoline intermediates, followed by the
Qiu-Yun Chen et al.
Colloids and surfaces. B, Biointerfaces, 114, 158-163 (2013-11-05)
Multifunctional silica nano-vehicles (SiO2@indol-IL) and magnetic nanoparticles (Fe3O4@indol-IL) were constructed through the Schiff bases condensation of indole-3-carboxaldehyde and 4-acetyl-N-allyl pyridinium chloride (ILs) with the amine groups of silica and magnetic nanoparticles. SiO2@indol-IL can inhibit the proliferation of HepG-2 cells in
Deepa Sinha et al.
European journal of medicinal chemistry, 43(1), 160-165 (2007-05-29)
Eight novel heterocyclic Schiff bases derived from the condensation reactions of indole 3-carboxaldehyde with different l-amino acids (histidine, glutamic acid, aspartic acid, leucine, valine) as well as with some aminophenols, have been synthesized and characterized by various spectroscopic methods (IR

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service