Skip to Content
Merck
All Photos(2)

Key Documents

795283

Sigma-Aldrich

Poly(2-methyl-2-oxazoline), hydroxy terminated

average Mn 5,000, PDI <1.3

Synonym(s):

POx 5kDa, PMOx

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(C4H7NO)nOH
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

description

Tg: 75-78°C

Quality Level

form

lumps

mol wt

average Mn 5,000

PDI

<1.3

storage temp.

2-8°C

InChI

1S/C5H11NO2/c1-5(8)6(2)3-4-7/h7H,3-4H2,1-2H3

InChI key

OBSKGKGKPGOOTB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Poly(2-methyl-2-oxazoline) is a hydrophilic polymer that is commonly used in biomedical applications. This is a biocompatible polymer with one chain-end functionalized with an OH group. They are commonly used as an amorphous alternative to PEG′s.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Poly(2-oxazoline)'s POx in Biomedical Applications.
Luxenhofer, R, and Jordan, R.
Material Matters, 8 (3), 70-72 null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service