Skip to Content
Merck
All Photos(1)

Key Documents

366765

Sigma-Aldrich

Methyl 2-methyl-3-nitrobenzoate

97%

Synonym(s):

Methyl 3-nitro-o-toluate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)CO2CH3
CAS Number:
Molecular Weight:
195.17
Beilstein:
1964020
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

62-65 °C (lit.)

functional group

ester
nitro

SMILES string

COC(=O)c1cccc(c1C)[N+]([O-])=O

InChI

1S/C9H9NO4/c1-6-7(9(11)14-2)4-3-5-8(6)10(12)13/h3-5H,1-2H3

InChI key

CRZGFIMLHZTLGT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methyl 2-methyl-3-nitrobenzoate may be used in the synthesis of:
  • methyl indole-4-carboxylate
  • 5-aminoisoquinolin-1(2H)-one
  • 5-nitroisocoumarin
  • substituted nitrostyrene benzoic acids, via reaction with aromatic aldehydes in the presence of DBU in DMSO
  • 4-(hydroxymethyl)-1-tosylindole, via Batcho-Leimgruber modification of the Reissert indole synthesis
  • [2-(4-fluorophenyl)-1H-indol-4-yl]-1-pyrrolidinylmethanone

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Chemistry of Indoles. XII. A Facile Route to 5-Nitroisocoumarins and Methyl Indole-4-carboxylate.
<BIG>Masanori S, et al. </BIG>
Chemical & Pharmaceutical Bulletin, 29(1), 249-253 (1981)
M C McDonald et al.
British journal of pharmacology, 130(4), 843-850 (2000-06-24)
Poly (ADP-ribose) synthetase (PARP) is a nuclear enzyme activated by strand breaks in DNA, which are caused inter alia by reactive oxygen species (ROS). Here we report on (i) a new synthesis of a water-soluble and potent PARP inhibitor, 5-aminoisoquinolinone
Preparation of 2-arylindole-4-carboxylic amide derivatives.
Kuethe JT and Davies LW.
Tetrahedron, 62(49), 11381-11390 (2006)
Synthesis of 2-Arylindole-4-Carboxylic Amides:[2-(4-Fluorophenyl)-1H-Indol-4-YL]-1-Pyrrolidinylmethanone.
Kuethe JT and Beutner GL.
Organic Syntheses, 92-104 (2009)
A N Brubaker et al.
Journal of medicinal chemistry, 29(8), 1528-1531 (1986-08-01)
Three 7-methyl-1,4-dioxa-7-azaspiro[4.5]decanes that contained either the benzyl, 3-indolylmethyl, or 4-indolylmethyl group at the 6-position were synthesized via alkylation of the pyrrolidine enamine of the key intermediate, ethyl 3-oxopiperidine-1-carboxylate. The spirodecane derivatives were evaluated for in vivo central and peripheral dopamine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service