Skip to Content
Merck
All Photos(5)

Documents

320102

Sigma-Aldrich

Acetic anhydride

ReagentPlus®, ≥99%

Synonym(s):

Acetanhydride, Acetic acid anhydride, Acetyl acetate, Acetyl anhydride, Ethanoic anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CO)2O
CAS Number:
Molecular Weight:
102.09
Beilstein:
385737
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

vapor density

3.5 (vs air)

Quality Level

vapor pressure

10 mmHg ( 36 °C)
4 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

≥99%

form

liquid

autoignition temp.

629 °F

expl. lim.

10.3 %

refractive index

n20/D 1.390 (lit.)

bp

138-140 °C (lit.)

mp

−73 °C (lit.)

solubility

water: slightly soluble

density

1.08 g/mL (lit.)

SMILES string

CC(=O)OC(C)=O

InChI

1S/C4H6O3/c1-3(5)7-4(2)6/h1-2H3

InChI key

WFDIJRYMOXRFFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Acetic anhydride is a carboxylic acid anhydride commonly used as an acetylating agent for amines and alcohols.
Acetic anhydride participates in the Friedel-Craft′s intramolecular cyclization reaction of cyclopropyl ketones to afford the tetralones. It has been reported to promote the Knoevenagel condensation of various aldehydes and activated methylene compounds.

Application

Acetic anhydride may be used as a reactant to synthesize:
  • 3-acetoxybenzoic acid by reacting with 3-hydroxybenzoic acid
  • 6-dimethylamino-2,3-naphthalenedicarboxylic anhydride, a precursor to prepare 6-dimethylaminonaphthalimide

Acetic anhydride was used in the following studies:
  • Lipozyme TL IM (commercial immobilized lipase from Thermomyces lanuginosus) catalyzed acetylation of essential clove oil.
  • As acetyl donor to investigate the lipase catalyzed acetylation of nanofibrillated cellulose (NFC).
  • Synthesis of N-substituted pyrazolines.
  • Acetylation of Barnyardgrass (Echinochloa crus-galli) starch.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

120.2 °F - closed cup

Flash Point(C)

49 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Braulio Insuasty et al.
European journal of medicinal chemistry, 93, 401-413 (2015-03-01)
A new series of chalcones 5a-f were synthesized from caffeine-based aldehyde 3 and substituted acetophenones 4a-f. Treatment of compounds 5a-f with hydrazine hydrate led to pyrazolines 6a-f, and their subsequent reaction with acetic anhydride or formic acid afforded the corresponding
Semi-synthesis and proteasome inhibition of D-ring deoxy analogs of (-)-epigallocatechin gallate (EGCG), the active ingredient of green tea extract.
Huo C, et al.
Journal of Chemistry, 86(6), 495-502 (2008)
Josiane Bartz et al.
Food chemistry, 178, 236-242 (2015-02-24)
Barnyardgrass (Echinochloa crus-galli) is an invasive plant that is difficult to control and is found in abundance as part of the waste of the paddy industry. In this study, barnyardgrass starch was extracted and studied to obtain a novel starch
Yohei Ogiwara et al.
The Journal of organic chemistry, 80(6), 3101-3110 (2015-02-18)
The combination of a catalytic amount of InCl3 and acetic anhydride remarkably promotes the Knoevenagel condensation of a variety of aldehydes and activated methylene compounds. This catalytic system accommodates aromatic aldehydes containing a variety of electron-donating and -withdrawing groups, heteroaromatic
Mojca Božič et al.
Carbohydrate polymers, 125, 340-351 (2015-04-11)
The acetylation efficiency of nanofibrillated cellulose (NFC) with acetic anhydride as acetyl donor was studied using lipase from Aspergillus niger in a mixture of dimethyl sulphoxide (DMSO) and phosphate buffer solution at ambient conditions and in supercritical carbon dioxide (scCO2).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service