Skip to Content
Merck
All Photos(2)

Key Documents

137847

Sigma-Aldrich

2-Methyl-3-nitrobenzoic acid

99%

Synonym(s):

3-Nitro-o-toluic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)CO2H
CAS Number:
Molecular Weight:
181.15
Beilstein:
2050096
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

mp

182-184 °C (lit.)

SMILES string

Cc1c(cccc1[N+]([O-])=O)C(O)=O

InChI

1S/C8H7NO4/c1-5-6(8(10)11)3-2-4-7(5)9(12)13/h2-4H,1H3,(H,10,11)

InChI key

YPQAFWHSMWWPLX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

2-Methyl-3-nitrobenzoic acid was used as starting reagent in the synthesis of methyl 2-methyl-3-nitrobenzoate and 2,3-unsubstituted indoles.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of (2S)-2-amino-3-(1H-4-indolyl) propanoic acid, a novel tryptophan analog for structural modification of bioactive peptides.
Fauq AH, et al.
Tetrahedron Asymmetry, 9(23), 4127-4134 (1998)
Palladium-catalyzed coupling of 2-bromoanilines with vinylstannanes. A regiocontrolled synthesis of substituted indoles.
Krolski ME, et al.
The Journal of Organic Chemistry, 53(6), 1170-1176 (1988)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service