Skip to Content
Merck
All Photos(1)

Key Documents

356867

Sigma-Aldrich

Cobalt

foil, thickness 0.1 mm, 99.95% trace metals basis

Synonym(s):

Cobalt element, Cobalt-59

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Co
CAS Number:
Molecular Weight:
58.93
EC Number:
MDL number:
UNSPSC Code:
12141710
PubChem Substance ID:
NACRES:
NA.23

Assay

99.95% trace metals basis

form

foil

resistivity

6.24 μΩ-cm, 20°C

thickness

0.1 mm

bp

2900 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

SMILES string

[Co]

InChI

1S/Co

InChI key

GUTLYIVDDKVIGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Quantity

2.2 g = 50 × 50 mm; 8.8 g = 100 × 100 mm

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Repr. 1A - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Perconti et al.
Journal of biological regulators and homeostatic agents, 27(2), 443-454 (2013-07-09)
Size-dependent characteristics of novel engineered nanomaterials might result in unforeseen biological responses and toxicity. To address this issue, we used cDNA microarray analysis (13443 genes) coupled with bioinformatics and functional gene annotation studies to investigate the transcriptional profiles of Balb/3T3
Heather De Bari et al.
Acta crystallographica. Section F, Structural biology and crystallization communications, 69(Pt 3), 228-236 (2013-03-23)
The X-ray crystal structure of ribosome hibernation promoting factor (HPF) from Vibrio cholerae is presented at 2.0 Å resolution. The crystal was phased by two-wavelength MAD using cocrystallized cobalt. The asymmetric unit contained two molecules of HPF linked by four Co
Jeannette Ø Penny et al.
Acta orthopaedica, 84(2), 130-137 (2013-04-20)
Wear particles from metal-on-metal arthroplasties are under suspicion for adverse effects both locally and systemically, and the DePuy ASR Hip Resurfacing System (RHA) has above-average failure rates. We compared lymphocyte counts in RHA and total hip arthroplasty (THA) and investigated
H John Cooper et al.
The Journal of bone and joint surgery. American volume, 95(10), 865-872 (2013-05-17)
Femoral stems with dual-taper modularity were introduced to allow additional options for hip-center restoration independent of femoral fixation in total hip arthroplasty. Despite the increasing availability and use of these femoral stems, concerns exist about potential complications arising from the
Carmen Gherasim et al.
The Journal of biological chemistry, 288(19), 13186-13193 (2013-03-30)
The reactivity of the cobalt-carbon bond in cobalamins is the key to their chemical versatility, supporting both methyl transfer and isomerization reactions. During evolution of higher eukaryotes that utilize vitamin B12, the high reactivity of the cofactor coupled with its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service