Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Cobalt
Synonym(s):
Cobalt
Empirical Formula (Hill Notation):
Co
CAS Number:
7440-48-4
Molecular Weight:
58.93
Products
Product Category
metals (1)
Brand
Sigma-Aldrich (10)
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Diameter (mm)
Length (mm)
Element
Quality Segment
Filter Products
Showing 1-20 of 201
Page 1 of 11
Page 1 of 11
Compare
Product Number
Description
Pricing
266639
Cobalt,
powder, 2 μm particle size, 99.8% trace metals basis
Expand
View Pricing
266647
Cobalt,
powder, <150 μm, ≥99.9% trace metals basis
Expand
View Pricing
203076
Cobalt,
granular, 99.99% trace metals basis
Expand
View Pricing
697745
Cobalt,
Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Expand
View Pricing
266655
Cobalt,
pieces, 99.5% trace metals basis
Expand
View Pricing
356867
Cobalt,
foil, thickness 0.1 mm, 99.95% trace metals basis
Expand
View Pricing
398810
Cobalt,
rod, diam. 5.0 mm, 99.95% trace metals basis
Expand
View Pricing
266701
Cobalt,
wire, diam. 1.0 mm, 99.995% trace metals basis
Expand
View Pricing
GF76259593
Cobalt,
foil, 100x100mm, thickness 2.0mm, as rolled, 99.9%
Expand
View Pricing
GF56494187
Cobalt,
foil, 100x100mm, thickness 1.0mm, as rolled, 99.9%
Expand
View Pricing
356891
Cobalt,
foil, thickness 1.0 mm, 99.95% trace metals basis
Expand
View Pricing
GF01015300
Cobalt,
powder, max. particle size 840 micron, weight 20 g, high purity 99.99+%
Expand
View Pricing
GF97256559
Cobalt,
lump, 10 mm max. lump size, weight 20 g, high purity 99.99+%
Expand
View Pricing
GF06340597
Cobalt,
powder, max. particle size 45 micron, weight 50 g, purity 99.8%
Expand
View Pricing
GF15401135
Cobalt,
foil, light tested, 25x25mm, thickness 0.01mm, 99.9%
Expand
View Pricing
326380
Cobalt,
foil, thickness 0.1 mm, ≥99.99%
Expand
View Pricing
GF87130728
Cobalt,
wire reel, 0.5m, diameter 0.05mm, hard, 99.9%
Expand
View Pricing
GF02492995
Cobalt,
rod, 2.0 mm diameter, length 2 mm, high purity 99.99+%
Expand
View Pricing
GF94243080
Cobalt,
foil, 8mm disks, thickness 0.01mm, 99.9%
Expand
View Pricing
GF63256743
Cobalt,
powder, 150 max. part. size (micron), weight 500 g, min. particle size 50 micron
Expand
View Pricing
Page 1 of 11
Page 1 of 11