Skip to Content
Merck
All Photos(2)

Key Documents

282863

Sigma-Aldrich

Graphite

powder, <20 μm, synthetic

Synonym(s):

carbon allotrope

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

powder

mol wt

Mw 12.011 g/mol

composition

C

particle size

<20 μm

mp

3652-3697 °C (lit.)

density

2.26 g/cm3 (lit.)

application(s)

battery manufacturing

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Graphite is an allotrope of carbon. Graphite is a nonmetallic showing high electrical conductivity. Synthetic graphite can be prepared by heating carbonaceous precursor in an inert atmosphere.
Graphite is the most stable allotrope of carbon under standard conditions.The only nonmetallic conductor of electricity. Graphite powder is used as an additive to alter the magnetorheology and electrical conductivity of magnetorheological elastomers (MREs).

Application

Graphite powder has been used:
  • in the synthesis of single sheet graphene nanoflakes
  • in the preparation of graphite based magnetorheological elastomers
  • free standing film like cantilever sensor in piezoelectric device
  • to study its use as a catalyst for wet peroxide oxidation.

Oxidation of carbon dust was studied.
Synthesis of graphite based MR elastomers have been reported. It may be used in the preparation of graphite oxide and anode in lithium ion battery.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Microstructure and magnetorheology of graphite based MR elastomers.
Tian TF, et al.
Rheologica Acta, 50, 825-836 (2011)
Ruben Sanchez-Hidalgo et al.
Polymers, 11(3) (2019-04-10)
Multifunctional elastomer nanocomposites have been applied in several high-tech fields. The design of materials with tailored properties capable of tuning their performance is a topical challenge. Here, we demonstrate that it is possible to modulate the mechanical and transport properties
Effect of mechanical milling of graphite powder on lithium intercalation properties
Natarajan C, et al.
The Journal of Practical Nursing, 92(1), 187-192 (2001)
Sensing capabilities of graphite based MR elastomers
Tian TF, et al.
Smart Materials and Structures, 20(2) (2011)
Layer-by-Layer Assembly of Ultrathin Composite Films from Micron-Sized Graphite Oxide Sheets and Polycations
Kovtyukhova NI, et al.
Chemistry of Materials, 11, 771-778 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service