Skip to Content
Merck
All Photos(3)

Key Documents

D3506

Sigma-Aldrich

Sodium diethyldithiocarbamate trihydrate

Synonym(s):

Cupral, Diethyldithiocarbamic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NCSSNa · 3H2O
CAS Number:
Molecular Weight:
225.31
Beilstein:
3920507
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

vapor density

5.9 (vs air)

Quality Level

Assay

≥97% (perchloric acid titration)

form

powder or crystals

mp

95-98.5 °C (lit.)

solubility

water: 1 g/10 mL, slightly hazy

storage temp.

room temp

SMILES string

[H]O[H].[H]O[H].[H]O[H].CCN(CC)C(=S)S[Na]

InChI

1S/C5H11NS2.Na.3H2O/c1-3-6(4-2)5(7)8;;;;/h3-4H2,1-2H3,(H,7,8);;3*1H2/q;+1;;;/p-1

InChI key

WWGXHTXOZKVJDN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Spin trap used in conjunction with Fe2+ to detect NO in brain, kidney, liver, and other tissues. Simultaneous direct measurement of NO* and pO2 has been reported. A copper chelator, it has been used to quantitate copper in biological materials by ESR and is an inhibitor of superoxide dismutase, ascorbate oxidase, and other enzymes.

Biochem/physiol Actions

Inhibits induction of macrophage nitric oxide synthase.

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Dopamine, Norepinephrine and Epinephrine Synthesis page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Caution

May form insoluble oxidation products in storage. Sensitivity in colorimetric copper determination not usually affected.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Suzuki et al.
Free radical research, 28(3), 293-299 (1998-08-04)
To detect nitric oxide (NO) in the rat brain during lipopolysaccharide (LPS)-induced sepsis, electron paramagnetic resonance (EPR) was employed with the NO trapping technique, using an iron and N,N-diethyldithiocarbamate (DETC) complex. An X-band (about 9.5 GHz) EPR system detected a
Tianhui Liu et al.
British journal of pharmacology, 175(24), 4480-4495 (2018-09-29)
Diethyldithiocarbamate (DDC) is a major metabolite of disulfiram that is a potential drug for alcoholism treatment. In the present study, we attempted to explore the possible effect of DDC on non-alcoholic fatty liver disease (NAFLD) and related fibrosis in vivo.
Jonathan H Pérez et al.
The Journal of experimental biology, 223(Pt 1) (2019-12-05)
The hypothalamic-pituitary-adrenal (HPA) axis is under complex regulatory control at multiple levels. Enzymatic regulation plays an important role in both circulating levels of glucocorticoids and target tissue exposure. Three key enzyme pathways are responsible for the immediate control of glucocorticoids.
Muriel Cuendet et al.
Nature protocols, 1(4), 1915-1921 (2007-05-10)
The cyclooxygenase (COX) reaction can be monitored by measurement of oxygen consumption, peroxidase co-substrate oxidation or prostaglandin (PG) detection. This protocol describes a procedure measuring cyclooxygenase activity by quantifying PGE2 produced by enzymatic conversion of arachidonic acid, in the presence
Gaurav Agrahari et al.
The Journal of investigative dermatology, 140(3), 656-665 (2019-08-30)
The expressions of LL-37 and KLK-5 were found to be altered in various dermatoses, including atopic dermatitis, psoriasis, and rosacea. However, the downstream inflammatory effect of LL-37 and KLK-5 is not as well studied. In addition, there is little high-quality

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service