326046
Yttrium(III) acetate hydrate
99.9% metals basis
Synonym(s):
Yttrium triacetate
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
(CH3CO2)3Y · xH2O
CAS Number:
Molecular Weight:
266.04 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Quality Level
Assay
99.9% metals basis
form
powder
reaction suitability
core: yttrium
reagent type: catalyst
SMILES string
O.CC(=O)O[Y](OC(C)=O)OC(C)=O
InChI
1S/3C2H4O2.H2O.Y/c3*1-2(3)4;;/h3*1H3,(H,3,4);1H2;/q;;;;+3/p-3
InChI key
JRKVGRAQLBXGQB-UHFFFAOYSA-K
Application
- Optical study of Yttrium oxide doped with zinc prepared by simple methods: The research investigates the optical properties of yttrium oxide doped with zinc, using yttrium acetate hydrate in the synthesis process. (Bhavani, Ganesan, 2015).
- Thermal decomposition of yttrium propionate: film and powder: This paper details the thermal decomposition behavior of yttrium propionate, a compound related to yttrium acetate, providing insights into decomposition mechanisms and thermal stability. (Rasi et al., 2018).
- In Situ Ternary Adduct Formation of Yttrium Polyaminocarboxylates Leads to Small Molecule Capture and Activation: This research investigates the formation of ternary adducts with yttrium complexes, using acetate and other ligands, demonstrating potential applications in small molecule activation. (Tickner et al., 2022).
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Daifeng Li et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 6(23), 1902042-1902042 (2019-12-14)
Recently, various second near-infrared window (NIR-II, 1000-1700 nm) fluorophores have been synthesized for in vivo imaging with nonradiation, high resolution, and low autofluorescence. However, most of the NIR-II fluorophores, especially inorganic nanoprobes, are mainly retained in the reticuloendothelial system (RES)
Huihui Wang et al.
Talanta, 197, 558-566 (2019-02-18)
A selective, simple and environment-friendly type fluorescence sensing platform between upconversion nanoparticles (UCNPs) and melanin-like polymer utilizing the photoinduced electron-transfer (PET) mechanism was developed. The effective fluorescence quenching of UCNPs by melanin-like polymers could detect tyramine and tyrosinase (TYR) activity
Qianqian Su et al.
Frontiers in chemistry, 8, 836-836 (2020-10-24)
Lanthanide-based upconversion nanoparticles can convert low-energy excitation to high-energy emission. The self-assembled upconversion nanoparticles with unique structures have considerable promise in sensors and optical devices due to intriguing properties. However, the assembly of isotropic nanocrystals into anisotropic structures is a
Wei Kong et al.
Inorganic chemistry, 56(2), 872-877 (2017-01-06)
Lanthanide-doped upconversion nanoparticles with a suitable surface coating are appealing for biomedical applications. Because high-quality upconversion nanoparticles are typically prepared in an organic solvent and passivated by hydrophobic oleate ligands, a convenient and reliable method for the surface modification of
Tianying Sun et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 17(5), 766-770 (2015-10-01)
Surface coating is a commonly used strategy to enhance upconversion emissions by shielding the luminescent core from surface quenching. In this work, we provide insights into the effect of surface coating on upconversion by investigating NaYF4 :Yb/Er nanoparticles and the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service