Skip to Content
Merck
All Photos(3)

Key Documents

464317

Sigma-Aldrich

Yttrium(III) chloride hexahydrate

99.99% trace metals basis

Synonym(s):

Yttrium trichloride hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
YCl3 · 6H2O
CAS Number:
Molecular Weight:
303.36
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder

Quality Level

Assay

99.99% trace metals basis

form

powder

impurities

≤150.0 ppm Trace Rare Earth Analysis

mp

100 °C (dec.) (lit.)

density

2.18 g/mL at 25 °C (lit.)

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].Cl[Y](Cl)Cl

InChI

1S/3ClH.6H2O.Y/h3*1H;6*1H2;/q;;;;;;;;;+3/p-3

InChI key

IINACGXCEZNYTF-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Yttrium(III) chloride hexahydrate can be used:
  • As a precursor to synthesize Y2O3–MgO nanocomposites via sol-gel processing.
  • To prepare finely dispersed yttrium-aluminum garnet (YAG) powder for laser ceramics.
  • As an additive to enhance the catalytic performance of activated carbon-supported Au catalyst (Au/AC).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sayed M Saleh et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 229, 117971-117971 (2020-01-19)
We present a new ratiometric and colorimetric optical sensor film for detection one of the most prevalent pesticide metribuzin. The detection proceeds within the low concentration range between 0 and 1.5 μM. The optical film is based on (a) near infrared
Yttrium chloride-modified Au/AC catalysts for acetylene hydrochlorination with improved activity and stability
Jinhuo Ke, et al.},
Journal of Rare Earths, 35, 1083-1091 (2017)
The influence of oxygen vacancy on the optical transmission of an yttria?magnesia nanocomposite
DongTao Jiang and Amiya K. Mukherjee
Scr. Mater., 64, 1095-1097 (2011)
Simone De Camillis et al.
Nanoscale, 12(39), 20347-20355 (2020-10-03)
Upconversion nanoparticles (UCNPs) exhibit unique optical properties such as photo-emission stability, large anti-Stokes shift, and long excited-state lifetimes, allowing significant advances in a broad range of applications from biomedical sensing to super-resolution microscopy. In recent years, progress on nanoparticle synthesis
Libing Fu et al.
Nanoscale, 9(36), 13683-13692 (2017-09-07)
There is considerable interest in developing diagnostic nanotools for early detection and delivery of various therapeutic agents for treatment of neurodegenerative diseases. However, a key challenge remains in the selection of suitable surfaces to overcome the nano-bio interface issue, namely

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service