326011
Ytterbium(III) acetate tetrahydrate
99.9% trace metals basis
Synonym(s):
Ytterbium(3+) triacetate tetrahydrate
About This Item
Recommended Products
Assay
99.9% trace metals basis
form
powder and chunks
reaction suitability
core: ytterbium
reagent type: catalyst
SMILES string
O.O.O.O.CC(=O)O[Yb](OC(C)=O)OC(C)=O
InChI
1S/3C2H4O2.4H2O.Yb/c3*1-2(3)4;;;;;/h3*1H3,(H,3,4);4*1H2;/q;;;;;;;+3/p-3
InChI key
ALJCLXGXZFZGEM-UHFFFAOYSA-K
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service