Skip to Content
Merck
All Photos(1)

Key Documents

325570

Sigma-Aldrich

Erbium(III) acetate hydrate

99.9% trace metals basis

Synonym(s):

Erbium triacetate hydrate, Erbium(3+) triacetate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CO2)3Er · xH2O
CAS Number:
Molecular Weight:
344.39 (anhydrous basis)
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace metals basis

form

crystalline powder
granular
powder, crystals or chunks

composition

Degree of hydration, ≤4

reaction suitability

core: erbium
reagent type: catalyst

SMILES string

O.O.O.O.CC(=O)O[Er](OC(C)=O)OC(C)=O

InChI

1S/3C2H4O2.Er.4H2O/c3*1-2(3)4;;;;;/h3*1H3,(H,3,4);;4*1H2/q;;;+3;;;;/p-3

InChI key

ZZGXSXSWDGRCAZ-UHFFFAOYSA-K

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuai Zha et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 8(5), e2002919-e2002919 (2021-03-16)
Using Epstein-Barr virus (EBV)-induced cancer cells and HeLa cells as a comparative study model, a novel and safe dual-EBV-oncoproteins-targeting pH-responsive peptide engineering, coating, and guiding approach to achieve precision targeting and treatment strategy against EBV-associated cancers is introduced. Individual functional
Lam Thi Kieu Giang et al.
Scientific reports, 10(1), 14672-14672 (2020-09-06)
This paper presents the use of soft template method to synthesize core and core-shell up-converting nanoparticles usefull for temperature sensing applications. Based on the stock solutions of core β-NaYF4:Er3+,Yb3+ nanoparticles and involving soft template method without any additional process of
Sanjeewa D Ranathunga et al.
Journal of dairy science, 102(9), 7980-7996 (2019-07-01)
The objective of this study was to investigate the effects of concentrations of forages and corn distillers dried grains with solubles (DDGS) on ruminal fermentation, ruminal kinetics, and total-tract digestibility of nutrients in lactating dairy cows. Four lactating Holstein cows
Aleksandra Kiseleva et al.
ACS applied materials & interfaces, 11(26), 22962-22972 (2019-06-30)
Spider silk is a natural material possessing unique properties such as biocompatibility, regenerative and antimicrobial activity, and biodegradability. It is broadly considered an attractive matrix for tissue regeneration applications. Optical monitoring and potential control over tissue regrowth are attractive tools
Yanxiao Ao et al.
ACS nano, 13(3), 3373-3386 (2019-01-27)
Near-infrared (NIR) light penetrates tissue deeply, but its application to motor behavior stimulation has been limited by the lack of known genetic NIR light-responsive sensors. We designed and synthesized a Yb3+/Er3+/Ca2+-based lanthanide-doped upconversion nanoparticle (UCNP) that effectively converts 808 nm

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service