Skip to Content
Merck
All Photos(2)

Documents

289256

Sigma-Aldrich

Erbium(III) chloride hexahydrate

99.9% trace metals basis

Synonym(s):

Erbium chloride hexahydrate, Erbium trichloride hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ErCl3·6H2O
CAS Number:
Molecular Weight:
381.71
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace metals basis

form

crystals and lumps

reaction suitability

reagent type: catalyst
core: erbium

impurities

≤1500.0 ppm Trace Rare Earth Analysis

SMILES string

O.O.O.O.O.O.Cl[Er](Cl)Cl

InChI

1S/3ClH.Er.6H2O/h3*1H;;6*1H2/q;;;+3;;;;;;/p-3

InChI key

UJBPGOAZQSYXNT-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Erbium(III) chloride hexahydrate may be used to prepare upconversion nanocrystal coated silica (UCNP@mSiO2) nanocomposite. The product may be used as a precursor for Er during the doping Y2O3 nanomaterials, which may be useful in bioimaging.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Controlled synthesis of uniform and monodisperse upconversion core/mesoporous silica shell nanocomposites for bimodal imaging.
Liu J, et al.
Chemistry?A European Journal , 18(8), 2335-2341 (2012)
Rare-earth-doped and codoped Y 2O 3 nanomaterials as potential bioimaging probes.
Das GK and Tan TTY
The Journal of Physical Chemistry C, 112(30), 11211-11217 (2008)
Xiangtong Zhang et al.
Journal of materials chemistry. C, 6(37), 10101-10105 (2018-12-07)
Lead halide perovskite nanocrystals (NCs) exhibit excellent tunable emissions covering the entire visible spectral region, but they do not emit near-infrared (NIR) light. We synthesized rare earth element doped CsPbCl3 NCs for NIR emission. The Yb3+ doped CsPbCl3 NCs emitted
Hongjun Zhuang et al.
Nanoscale, 12(18), 10216-10225 (2020-05-02)
Because of the low tissue penetration depth and poor photostability of organic cyanine dye, in addition to environmental interference, it is a great challenge to monitor the degree of drug-induced hepatotoxicity by the in vivo detection of peroxynitrite (ONOO-). Herein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service