A59522
2-Aminoindan hydrochloride
98%
Synonym(s):
2-Indanamine hydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C9H11N · HCl
CAS Number:
Molecular Weight:
169.65
Beilstein:
3913700
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
crystals
mp
245-247 °C (lit.)
SMILES string
Cl[H].NC1Cc2ccccc2C1
InChI
1S/C9H11N.ClH/c10-9-5-7-3-1-2-4-8(7)6-9;/h1-4,9H,5-6,10H2;1H
InChI key
XEHNLVMHWYPNEQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Prepared from 2-indanols via azide displacement and subsequent hydrogenation.
Signal Word
Warning
Hazard Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Loanda R Cumba et al.
The Analyst, 140(16), 5536-5545 (2015-06-24)
"Synthacaine" is a New Psychoactive Substance which is, due to its inherent psychoactive properties, reported to imitate the effects of cocaine and is therefore consequently branded as "legal cocaine". The only analytical approach reported to date for the sensing of
W Dimpfel et al.
BMC pharmacology, 11, 2-2 (2011-02-23)
Rasagiline, a new drug developed to treat Parkinson's disease, is known to inhibit monoamine oxidase B. However, its metabolite R-(-)-aminoindan does not show this kind of activity. The present series of in vitro experiments using the rat hippocampal slice preparation
S X Ma et al.
The Journal of pharmacology and experimental therapeutics, 256(2), 751-756 (1991-02-01)
Dopaminergic structure-activity relationships of 2-aminoindans were evaluated for their ability to inhibit responses to stimulation of cardioaccelerator nerves in cats. The major observations were as follows: 1) Unsubstituted di-n-propyl- and diethyl 2-aminoindan derivatives do not inhibit responses to stimulation of
H Wikström et al.
Journal of medicinal chemistry, 30(7), 1115-1120 (1987-07-01)
Conformational analyses on differently substituted 2-aminoindans of significant pharmacological interest were carried out by the molecular mechanics method (MM2). An X-ray structure of (R)-4-methoxy-2-aminoindan has shown the ammonium nitrogen [-)-D-tartaric acid salt) in an axial position. From comparison with other
S P Arnerić et al.
Archives internationales de pharmacodynamie et de therapie, 258(1), 84-99 (1982-07-01)
The ability of 2-aminotetralins (2-ATs), 2-aminoindanes (2-AIs), morphine (M) and clonidine (CLON) to alter neuroeffector transmission was studied on field-stimulated (FS) guinea-pig ilea (GPI). The activity of these compounds to inhibit K+, histamine (H), actylcholine (ACh), nicotine (Nic) and serotonin
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service