Skip to Content
Merck
All Photos(1)

Key Documents

12635

Sigma-Aldrich

Benzenesulfonic acid

98.0% (T)

Synonym(s):

Phenylsulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SO3H
CAS Number:
Molecular Weight:
158.18
Beilstein:
742513
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

98.0% (T)

form

solid

impurities

≤1.0% water

functional group

sulfonic acid

SMILES string

OS(=O)(=O)c1ccccc1

InChI

1S/C6H6O3S/c7-10(8,9)6-4-2-1-3-5-6/h1-5H,(H,7,8,9)

InChI key

SRSXLGNVWSONIS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzenesulfonic acid (BSA), also known as phenylsulfonic acid, is the simplest aromatic sulfonic acid. It is also used as a catalyst for Michael′s reactions and in the oxidative alkenylation process. [1]

Application

Benzenesulfonic acid is an aryl sulfonic acid that can be used:
  • As an oxidizing agent in the synthesis of Fe(III) benzenesulfonate. [2]
  • As a catalyst during the vapor-phase nitration of benzene. [3]
  • As a reagent in the synthesis of organic salts. [4]
  • To form an organic salt with 5,7-dimethyl-1,8-naphthyridine-2-amine, that can be a synthon for developing supramolecular structures.[4]
  • As a dopant for the polymerization of pyrrole to form poly(pyrrole), a conducting polymer useful in the development of flexible capacitors.[5]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Vapor-phase nitration of benzene over silica-supported benzenesulfonic acid catalyst
Eiichi S,et al.
Chemistry Letters (Jpn), 16, 2273-2274 (1987)
Vapour phase polymerisation of pyrrole induced by iron (III) alkylbenzenesulfonate salt oxidising agents
Priya S,et al.
Synthetic Metals, 158, 704-711 (2008)
Supramolecular salts of 5, 7-dimethyl-1, 8-naphthyridine-2-amine and acids through classical H-Bonds and other intermolecular interactions.
Dong L
Journal of Molecular Structure, 1153, 311-323 (2018)
Durable Flexible Supercapacitors Utilizing the Multifunctional Role of Ionic Liquids.
Lorenzo M
Energy Technology, 6(1), 196-204 (2018)
David Scholz et al.
ChemSusChem, 11(13), 2189-2201 (2018-05-08)
The deactivation pathways of sulfonated carbon catalysts prepared from different carbons were studied during the aqueous-phase hydrolysis of cellobiose under continuous-flow conditions. The sulfonation of carbon materials with a low degree of graphitization introduced sulfonic acid groups that are partially

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service