Skip to Content
Merck
All Photos(1)

Key Documents

302015

Sigma-Aldrich

5-Hexyn-1-ol

96%

Synonym(s):

1-Hexyn-6-ol, 1-Hydroxy-5-hexyne, 5-Hexynol, 5-Hexynyl alcohol, 6-Hydroxy-1-hexyne

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC≡C(CH2)4OH
CAS Number:
Molecular Weight:
98.14
Beilstein:
1739774
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

96%

form

liquid

refractive index

n20/D 1.450 (lit.)

bp

73-75 °C/15 mmHg (lit.)

density

0.89 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

OCCCCC#C

InChI

1S/C6H10O/c1-2-3-4-5-6-7/h1,7H,3-6H2

InChI key

GOQJMMHTSOQIEI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

5-Hexyn-1-ol was used in synthesis of:
  • cinnoline-fused cyclic enediyne[1]
  • llycopodium alkaloids, (+)-nankakurine A and (+)-nankakurine B[2]
  • 7-benzoyloxy-3-(2-nitrophenylseleno)-1,5-cyclodecadiyne[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryan A Altman et al.
The Journal of organic chemistry, 75(22), 7519-7534 (2010-10-21)
The first total syntheses of the Lycopodium alkaloids (+)-nankakurine A (2), (+)-nankakurine B (3), and the originally purported structure 1 of nankakurine A were accomplished. The syntheses of 2 and 3 feature a demanding intramolecular azomethine imine cycloaddition as the
Olga V Vinogradova et al.
The Journal of organic chemistry, 76(16), 6937-6941 (2011-07-02)
A short and efficient synthesis of cinnoline-fused cyclic enediyne is reported. Richter cyclization of o-(1,3-butadiynyl)phenyltriazene produced 3-alkynyl-4-bromocinnoline. The Sonogashira coupling of the latter with 5-hexyn-1-ol was employed for the introduction of a second acetylenic moiety. The crucial cyclization step was
M Michida et al.
Nucleic acids symposium series, (37)(37), 55-56 (1997-01-01)
7-Benzoyloxy-3-(2-nitrophenylseleno)-1,5-cyclodecadiyne (13) was prepared from 5-hexyn-1-ol and propargyl bromide via 10-iodo-7-(t-butyldiphenylsiloxy)-5,9-decadiynal (10). The facile cyclization of the acyclic precursor 10 would be rationalized in terms of "bulky group (TBDPSO) assisted conformational control". Oxidation of 13 by NaIO4, followed by thermal
Kun Liu et al.
Nucleic acids research, 48(13), 7356-7370 (2020-06-11)
To enable the optimal, biocompatible and non-destructive application of the highly useful copper (Cu+)-mediated alkyne-azide 'click' cycloaddition in water, we have isolated and characterized a 79-nucleotide DNA enzyme or DNAzyme, 'CLICK-17', that harnesses as low as sub-micromolar Cu+; or, surprisingly
Johanna E Grimm et al.
Journal of agricultural and food chemistry, 67(20), 5838-5846 (2019-05-06)
An aroma extract dilution analysis applied to the volatiles isolated from jackfruit ( Artocarpus heterophyllus Lam.) pulp by solvent extraction and solvent-assisted flavor evaporation resulted in the detection of 48 odorants with flavor dilution (FD) factors between 1 and ≥8192.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service