Skip to Content
Merck
All Photos(1)

Key Documents

202924

Sigma-Aldrich

Cadmium sulfate hydrate

≥99.995% trace metals basis

Synonym(s):

Sulfuric acid cadmium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CdSO4 · xH2O
CAS Number:
Molecular Weight:
208.47 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
≥99.995% trace metals basis
form:
powder and chunks

Quality Level

Assay

≥99.995% trace metals basis

form

powder and chunks

reaction suitability

reagent type: catalyst
core: cadmium

impurities

≤50.0 ppm Trace Metal Analysis

SMILES string

O.[Cd++].[O-]S([O-])(=O)=O

InChI

1S/Cd.H2O4S.H2O/c;1-5(2,3)4;/h;(H2,1,2,3,4);1H2/q+2;;/p-2

InChI key

VRFRVKRTIMIRNG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Applied in the formation of novel two-dimensional Cd-SCN coordination solids with unusual and tailorable, checkerboard- or herringbone-patterned structures these structures are important steps toward technologically useful materials.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 1B - Repr. 1B - STOT RE 1

Target Organs

Bone,Kidney,Respiratory system

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Manjushree et al.
Bioinorganic chemistry and applications, 2018, 6954951-6954951 (2018-07-28)
The binding interaction between bovine serum albumin (BSA) and sodium salt of risedronic acid (RSN) was studied by using the FT-IR (Fourier transform infrared), UV-Vis (ultraviolet-visible), fluorescence (emission and synchronous), CD (circular dichroism) spectrometric, and computational (molecular docking) techniques at
Akiko Honda et al.
Environmental toxicology and pharmacology, 52, 276-279 (2017-05-10)
We investigated the synergic effects of components of particulate matter with aerodynamic diameters ≤2.5μm (PM2.5) on airway inflammation. Co-exposure to cadmium (Cd) and 9,10-phenanthrenequinone (9,10-PQ) additively/synergistically increased pro-inflammatory responses in airway epithelial cells, whereas co-exposure to Cd and phenanthrene resulted
Katharina Ruppert et al.
Chemosphere, 181, 589-599 (2017-05-05)
Mollusks are known to be uniquely sensitive to a number of reproductive toxicants including some vertebrate endocrine disrupting chemicals. However, they have widely been ignored in environmental risk assessment procedures for chemicals. This study describes the validation of the Potamopyrgus
Hong Zhang et al.
Inorganic chemistry, 38(5), 886-892 (2001-10-24)
This paper reports the syntheses, characterization, and structures of two new two-dimensional cadmium-thiocyanate coordination solids [(12C4)(2)Cd][Cd(2)(SCN)(6)] (1) and [(12C4)(2)Cd][Cd(3)(SCN)(8)] (2), where 12C4 denotes 12-crown-4, C(8)H(16)O(4). 1 crystallizes in a monoclinic unit cell, space group C2/c with lattice parameters a =
Junsung Woo et al.
Experimental neurobiology, 28(1), 30-42 (2019-03-12)
The neuronal activity-dependent change in the manner in which light is absorbed or scattered in brain tissue is called the intrinsic optical signal (IOS), and provides label-free, minimally invasive, and high spatial (~100 µm) resolution imaging for visualizing neuronal activity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service