Skip to Content
Merck
All Photos(2)

Key Documents

202886

Sigma-Aldrich

Cadmium

shot, 3 mm, 99.999% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cd
CAS Number:
Molecular Weight:
112.41
Beilstein:
8137359
EC Number:
MDL number:
UNSPSC Code:
12141707
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.999% trace metals basis

form

shot

resistivity

7.27 μΩ-cm, 22°C

particle size

3 mm

bp

765 °C (lit.)

mp

320.9 °C (lit.)

density

8.65 g/mL at 25 °C (lit.)

SMILES string

[Cd]

InChI

1S/Cd

InChI key

BDOSMKKIYDKNTQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 2 - Repr. 2 - STOT RE 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Boldizsar Nagy et al.
Acta chimica Slovenica, 60(2), 263-273 (2013-07-25)
Biosorption of cadmium ions from synthetic aqueous solution using popular Romanian fir tree sawdust (Abies Alba) as biosorbent, was investigated in this work. Prior to its utilization the considered biomass was washed, dried and sieved without further chemical treatments. The
F Tomei et al.
Annali di igiene : medicina preventiva e di comunita, 25(4), 353-363 (2013-05-25)
The exposure to Cd has been linked to many disease and liver is the major target organ of Cd toxicity following acute or chronic exposure.No study has ever evaluated the association between liver function tests and exposure to Cd present
Gang Zhang et al.
Journal of environmental sciences (China), 25(1), 231-235 (2013-04-17)
The emission characteristics of polychlorinated dibenzo-p-dioxins and dibenzofurans (PCDD/Fs) and heavy metals were evaluated during co-combustion of sewage sludge with coal from a circulating fluidized bed incinerator. The stack gas, slag and fly ash samples were sampled and analyzed. The
Simona Ben-Haim et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 54(6), 873-879 (2013-04-13)
Myocardial perfusion imaging (MPI) is well established in the diagnosis and workup of patients with known or suspected coronary artery disease (CAD); however, it can underestimate the extent of obstructive CAD. Quantification of myocardial perfusion reserve with PET can assist
Shalini Jhanji et al.
Journal of environmental biology, 33(6), 1027-1032 (2013-06-08)
Experiments were carried out to study the effect of cadmium (Cd) and exogenous nitric oxide (NO) on growth, photosynthetic attributes, yield components and structural features of Brassica napus L. (cv. GSL 1). Cadmium in the growth medium at different levels

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service