Skip to Content
Merck
All Photos(1)

Documents

250627

Sigma-Aldrich

Chlorotriisopropoxytitanium(IV)

95%

Synonym(s):

Chlorotitanium(IV) triisopropoxide, Titanium(IV) chloride triisopropoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CHO]3TiCl
CAS Number:
Molecular Weight:
260.58
Beilstein:
4163222
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

form

liquid

reaction suitability

core: titanium
reagent type: catalyst

mp

35-40 °C

density

1.091 g/mL at 25 °C (lit.)

SMILES string

CC(C)O[Ti](Cl)(OC(C)C)OC(C)C

InChI

1S/3C3H7O.ClH.Ti/c3*1-3(2)4;;/h3*3H,1-2H3;1H;/q3*-1;;+4/p-1

InChI key

IFMWVBVPVXRZHE-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Useful additive for increased regio- and stereocontrol in aldol reactions.

Other Notes

Storage below 25 °C may cause crystallization. Moving container to a warm location should remelt the solid (mp ~35 °C). Recrystallization occurs slowly. Thus, the liquid may be transferred using syringe techniques.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - Skin Corr. 1B

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 34, 677-677 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service