Skip to Content
Merck
All Photos(1)

Key Documents

333484

Sigma-Aldrich

Titanium(IV) 2-ethylhexyloxide

95%

Synonym(s):

Tetra(2-ethylhexyl)titanate, Tetrakis(2-ethylhexyl) orthotitanate, TYZOR® TOT organic titanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ti[OCH2CH(C2H5)(CH2)3CH3]4
CAS Number:
Molecular Weight:
564.75
Beilstein:
3692023
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

95%

form

liquid
liquid

reaction suitability

core: titanium
reagent type: catalyst

refractive index

n20/D 1.479 (lit.)

bp

248-249 °C/11 mmHg (lit.)

density

0.927 g/mL at 25 °C (lit.)

SMILES string

CCCCC(CC)CO[Ti](OCC(CC)CCCC)(OCC(CC)CCCC)OCC(CC)CCCC

InChI

1S/4C8H17O.Ti/c4*1-3-5-6-8(4-2)7-9;/h4*8H,3-7H2,1-2H3;/q4*-1;+4

InChI key

KTXWGMUMDPYXNN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

TYZOR is a registered trademark of E. I. du Pont de Nemours and Company

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li-Min Wang et al.
The journal of physical chemistry. B, 109(22), 11091-11094 (2006-07-21)
Many hydrogen-bonded liquids, especially glass-forming cases, display a dielectric relaxation behavior that differs qualitatively from that of other simple liquids. The majority of models aimed at explaining this unusual dielectric behavior associate the prominent Debye process with structural relaxation, viscous
Gunilla Wieslander et al.
Archives of environmental & occupational health, 65(1), 3-11 (2010-02-12)
The authors evaluated changes of symptoms and biomarkers in health care staff (N = 18) for people with different physical dysfunctions and similarly in an external office control group in a nondamp building (N = 15). The first workplace had
Adam R Grochowalski et al.
Biodegradation, 18(3), 283-293 (2006-11-03)
The biodegradation of plasticizers has been previously shown to result in the accumulation of metabolites that are more toxic than the initial compound. The present work shows that the pattern of degradation of di-2-ethylhexyl adipate by Bacillus subtilis can be
Kiyoshi Sakai et al.
Journal of environmental monitoring : JEM, 11(11), 2068-2076 (2009-11-06)
2-Ethyl-1-hexanol (2E1H) is a possible causative chemical for sick building symptoms; however, thus far, we do not have a clear understanding of the indoor air pollution levels caused by it. In this study, first, airborne 2E1H concentrations were measured during
D Fragiadakis et al.
The Journal of chemical physics, 132(14), 144505-144505 (2010-04-22)
The dielectric spectra of most simple liquids are characterized by two relaxation processes: (i) the alpha-process, an intense, broad non-Debye relaxation with a non-Arrhenius temperature dependence and (ii) a beta process, evident mainly below the glass transition and having nearly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service