Skip to Content
Merck
All Photos(3)

Documents

246948

Sigma-Aldrich

(R)-(+)-1,1′-Bi(2-naphthol)

99%

Synonym(s):

(+)-2,2′-Dihydroxy-1,1′-dinaphthyl, (R)-(+)-1,1′-Binaphthalene-2,2′-diol, (R)-BINOL

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC10H6C10H6OH
CAS Number:
Molecular Weight:
286.32
Beilstein:
3616837
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
EC Index Number:
606-048-4
NACRES:
NA.22

Assay

99%

form

solid

optical activity

[α]21/D +34°, c = 1 in THF

optical purity

ee: 99% (HPLC)

mp

208-210 °C (lit.)

SMILES string

Oc1ccc2ccccc2c1-c3c(O)ccc4ccccc34

InChI

1S/C20H14O2/c21-17-11-9-13-5-1-3-7-15(13)19(17)20-16-8-4-2-6-14(16)10-12-18(20)22/h1-12,21-22H

InChI key

PPTXVXKCQZKFBN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

A chiral auxiliary used in the catalytic asymmetric oxidation of sulfides to sulfoxides. Chiral lanthanide triflates formed from binaphthol serve as catalysts for asymmetric Diels-Alder reactions. Derivatives of binaphthol have recently found use in asymmetric Claisen rearrangements and asymmetric epoxidations. The lithium aluminum hydride derivative of these diols (BINAP-H) has been used extensively for the reduction of ketones.
Chiral binapthol imminium salt precursor. Salts were used for an asymmetric epoxidation of olefins.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Asymmetric Diels-Alder reactions catalyzed by chiral lanthanide (III) trifluoromethanesulfonates. Unique structure of the triflate and stereoselective synthesis of both enantiomers using a single chiral source and a choice of achiral ligands.
S. Kobayashi et al.
Tetrahedron, 50(40), 11623-11623 (1994)
Philip C Bulman Page et al.
The Journal of organic chemistry, 72(12), 4424-4430 (2007-05-18)
A series of binaphthalene-fused azepinium salts has been generated and used as organocatalysts in the asymmetric epoxidation of unfunctionalized alkenes, giving rise to ees of up to 84%.
Asymmetric aza-Diels-Alder reaction catalyzed by boron reagent: Effect of biphenol and binaphthol ligand.
Hattori, K. Yamamoto, H.
Synlett, 1993(02), 129-129 (1993)
Catalytic asymmetric oxidation of sulfides to sulfoxides with tert-butyl hydroperoxide using binaphthol as a chiral auxiliary.
Komatsu N, et al.
The Journal of Organic Chemistry, 58, 4529-4529 (1993)
Catalytic asymmetric epoxidation with (Salen) manganese (III) complex bearing binaphthyl groups of axial chirality.
Sasaki, H. et al.
Synlett, 1993(04), 300-300 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service