Skip to Content
Merck
All Photos(2)

Key Documents

14010

Sigma-Aldrich

Titanium(III) chloride solution

10-15% TiCl3 basis

Synonym(s):

Titanium trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiCl3
CAS Number:
Molecular Weight:
154.23
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

Quality Level

reaction suitability

core: titanium
reagent type: catalyst

concentration

10-15% (TiCl3)
12% in hydrochloric acid

impurities

5-12% free acid (as HCl)

SMILES string

Cl[Ti](Cl)Cl

InChI

1S/3ClH.Ti/h3*1H;/q;;;+3/p-3

InChI key

YONPGGFAJWQGJC-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for hydrogen desorption and nitrogen-nitrogen bond cleavage

Reagent for photocatalyst synthesis

Precursor for preparation of nanostructures for nanoskeletons

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shigehite Isobe, et al
Applied Physics Letters, 99 (2011)
B Barrocas et al.
Chemosphere, 250, 126240-126240 (2020-03-03)
The unexpected incorporation of ionic Mn and Fe in the crystalline structure of titanate nanowires was accomplished when a contaminated a titanium source was used. The presence of Mn (8.1 mg L-1) and Fe (4.3 mg L-1) result in the production of a novel
Kevin R MacKenzie et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 153, 105488-105488 (2020-07-28)
Atomoxetine (ATX), a selective and potent inhibitor of the presynaptic norepinephrine transporter, is used mainly to treat attention-deficit hyperactivity disorder. Although multiple adverse effects associated with ATX have been reported including severe liver injuries, the mechanisms of ATX-related toxicity remain
D. Meroni, et al
Catalysis Today, 161, 169-174 (2011)
Shuxi Dai et al.
Nanoscale research letters, 5(11), 1829-1835 (2010-12-03)
Highly crystalline TiO(2) nanostructures were prepared through a facile inorganic acid-assisted hydrothermal treatment of hexagonal-structured assemblies of nanocrystalline titiania templated by cetyltrimethylammonium bromide (Hex-ncTiO(2)/CTAB Nanoskeleton) as starting materials. All samples were characterized by X-ray diffraction (XRD) and transmission electron microscopy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service