Skip to Content
Merck
All Photos(1)

Documents

302929

Sigma-Aldrich

Tetraethylammonium hydroxide solution

35 wt. % in H2O

Synonym(s):

TEAOH solution, TEA hydroxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)4N(OH)
CAS Number:
Molecular Weight:
147.26
Beilstein:
3912961
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

liquid

concentration

35 wt. % in H2O

refractive index

n20/D 1.404

density

1.023 g/mL at 25 °C

SMILES string

[OH-].CC[N+](CC)(CC)CC

InChI

1S/C8H20N.H2O/c1-5-9(6-2,7-3)8-4;/h5-8H2,1-4H3;1H2/q+1;/p-1

InChI key

LRGJRHZIDJQFCL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

TEAOH solution is a strong organic base used to adjust pH of the acidic mobile phase in chromatographic separation.

Application

Tetraethylammonium hydroxide solution (35 wt. % in H2O) may be used as an organic structure-directing agent (SDA)/template in the synthesis of mesoporous molecular sieves and zeolite samples.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of nanocrystalline zeolite beta in the absence of alkali metal cations.
Camblor MA
Studies in Surface Science and Catalysis, 105, 341-348 (1997)
Tin-containing zeolites are highly active catalysts for the isomerization of glucose in water.
Moliner M
Proceedings of the National Academy of Sciences of the USA, 107(14), 6164-6168 (2010)
Kyohei Ueno et al.
Membranes, 9(7) (2019-07-20)
Hydrophobic pure-silica *BEA-type zeolite membranes with large pores were prepared on tubular silica supports by hydrothermal synthesis using a secondary growth method and were applied to the separation of alcohol/water mixtures by pervaporation (PV), an alternative energy-efficient process for production
Quantification of the enantiomers of ofloxacin in biological fluids by high-performance liquid chromatography
Lehr KH, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 425, 153-161 (1988)
Matjaž Mazaj et al.
ChemSusChem, 13(8), 2089-2097 (2020-01-23)
Here, the facile synthesis of four-length-scaled (ultramicro-micro-meso-macroporous) hierarchically structured porous carbon nanocomposite by an emulsion-template strategy is reported. This previously unreported combination of zeolite nanocrystals embedded in the walls of microcellular carbon foams gives unique textural and structural properties, which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service