Skip to Content
Merck
All Photos(1)

Key Documents

668281

Sigma-Aldrich

1-Hydroxytetraphenyl-cyclopentadienyl(tetraphenyl-2,4-cyclopentadien-1-one)-μ-hydrotetracarbonyldiruthenium(II)

98%

Synonym(s):

2,4-Cyclopentadien-1-one, 2,3,4,5-tetraphenyl-, ruthenium complex, Shvo’s Catalyst, Tetracarbonyl-μ-hydro[(1,2,3,4,5-H)-1-hydroxylato-2,3,4,5-tetraphenyl-2,4-cyclopentadien-1-yl][(1,2,3,4,5-H)-1-hydroxy-2,3,4,5-tetraphenyl-2,4-cyclopentadien-1-yl]diruthenium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C62H42O6Ru2
CAS Number:
Molecular Weight:
1085.13
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

reaction suitability

core: ruthenium
reagent type: catalyst

mp

223-227 °C

SMILES string

[Ru][RuH].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].O[C]1[C]([C]([C]([C]1c2ccccc2)c3ccccc3)c4ccccc4)c5ccccc5.O=C6C(c7ccccc7)=C(c8ccccc8)C(c9ccccc9)=C6c%10ccccc%10

InChI

1S/C29H21O.C29H20O.4CO.2Ru.H/c2*30-29-27(23-17-9-3-10-18-23)25(21-13-5-1-6-14-21)26(22-15-7-2-8-16-22)28(29)24-19-11-4-12-20-24;4*1-2;;;/h1-20,30H;1-20H;;;;;;;

InChI key

PISMVXYUJBWZFB-UHFFFAOYSA-N

Application

Has found numerous applications as a versatile catalyst in organic synthesis, including the reduction of aldehydes and ketones to alcohols, bimolecular disproportionation reaction of aldehydes to esters, isomerization of allylic alcohols and oxidation of alcohols.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prabhakaran, R.
Synlett, 2048-2048 (2004)
Combination of enzymes and metal catalysts. A powerful approach in asymmetric catalysis.
Oscar Pàmies et al.
Chemical reviews, 103(8), 3247-3262 (2003-08-14)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service