Skip to Content
Merck
All Photos(1)

Key Documents

244759

Sigma-Aldrich

Titanium(IV) ethoxide

technical grade

Synonym(s):

Tetraethyl orthotitanate, Tetraethyl titanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ti(OC2H5)4
CAS Number:
Molecular Weight:
228.11
Beilstein:
3678992
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

form

liquid

composition

Ti, 19.0-21.0% gravimetric

reaction suitability

core: titanium
reagent type: catalyst

impurities

~20% Tetraisopropyl orthotitanate

refractive index

n20/D 1.505 (lit.)

bp

150-152 °C/10 mmHg (lit.)

density

1.088 g/mL at 25 °C (lit.)

SMILES string

CCO[Ti](OCC)(OCC)OCC

InChI

1S/4C2H5O.Ti/c4*1-2-3;/h4*2H2,1H3;/q4*-1;+4

InChI key

JMXKSZRRTHPKDL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Titanium ethoxide is used widely as a precursor for the synthesis of Ti4+ ions titania nanoparticles by various techniques such as :
  • in the sol-gel synthesis of free-standing ferroelectric lead zirconate titanate nanoparticles
  • spherical titania cores, produced either by hydrolysis of Ti(IV) ethoxide droplets in contact with water vapor or
  • by preadsorption of the titanium alkoxides into/onto the clay mineral platelets and their subsequent hydrolysis and calcination.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

107.6 °F - closed cup

Flash Point(C)

42 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of uniform spherical titania particles coated with polyurea by the aerosol technique.
Mayville FC, et al.
Journal of Colloid and Interface Science, 120(1), 135-139 (1987)
Two-photon polymerization of metal ions doped acrylate monomers and oligomers for three-dimensional structure fabrication
Duan XM, et al.
Thin Solid Films, 453, 518-521 (2004)
Improved synthesis of enantiopure sulfinimines (thiooxime s-oxides) from p-toluenesulfinamide and aldehydes and ketones
Davis FA, et al.
The Journal of Organic Chemistry, 64(4), 1403-1406 (1999)
Preparation and characterization of clay mineral intercalated titanium dioxide nanoparticles
Mogyorosi K, et al.
Langmuir, 19(7), 2938-2946 (2003)
Johan Karlsson et al.
International journal of nanomedicine, 10, 4425-4436 (2015-07-18)
The osseointegration capacity of bone-anchoring implants can be improved by the use of drugs that are administrated by an inbuilt drug delivery system. However, to attain superior control of drug delivery and to have the ability to administer drugs of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service