Skip to Content
Merck
All Photos(1)

Documents

272000

Sigma-Aldrich

2-Imidazolecarboxaldehyde

97%

Synonym(s):

1H-Imidazole-2-carbaldehyde, 2-Formylimidazole, 2-Imidazolecarbaldehyde, 2-Imidazolylformaldehyde, Imidazole-2-formaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4N2O
CAS Number:
Molecular Weight:
96.09
Beilstein:
4371302
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

209 °C (dec.) (lit.)

SMILES string

[H]C(=O)c1ncc[nH]1

InChI

1S/C4H4N2O/c7-3-4-5-1-2-6-4/h1-3H,(H,5,6)

InChI key

XYHKNCXZYYTLRG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Imidazolecarboxaldehyde was used in the preparation of tridentate Schiff-base carboxylate-containing ligands by undergoing condensation reaction with amino acids β-alanine and 2-aminobenzoic acid. It was also used in a study of the imidazole-directed allylation of aldimines.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tohid Taghizadeh et al.
International journal of biological macromolecules, 166, 1301-1311 (2020-11-09)
A zeolitic imidazolate framework (ZIF-90) has been synthesized through solvothermal method. The structure was characterized by means of FT-IR spectroscopy, X-ray diffraction, thermogravimetric analysis (TGA), and scanning electron microscopy (SEM)/energy dispersive X-ray spectroscopy (EDS). The synthesized ZIF-90 was applied as
Rahul Sharma et al.
Environmental toxicology and pharmacology, 80, 103454-103454 (2020-07-10)
The present armamentarium of commercially available antidotes provides limited protection against the neurological effects of organophosphate exposure. Hence, there is an urgent need to design and develop molecules that can protect and reactivate inhibited-AChE in the central nervous system. Some
E Colacio et al.
Inorganic chemistry, 39(13), 2882-2890 (2001-03-10)
Tridentate Schiff-base carboxylate-containing ligands, derived from the condensation of 2-imidazolecarboxaldehyde with the amino acids beta-alanine (H2L1) and 2-aminobenzoic acid (H2L5) and the condensation of 2-pyridinecarboxaldehyde with beta-alanine (HL2), D,L-3-aminobutyric acid (HL3), and 4-aminobutyric acid (HL4), react with copper(II) perchlorate to
Ruirui Li et al.
Small (Weinheim an der Bergstrasse, Germany), 15(52), e1904866-e1904866 (2019-11-30)
The confinement effect in asymmetrical biological ion channels makes the state of molecules and ions differs from that in the external environment, and the mass transfer confined in the biological ion channels is in a single strand form. Herein, an
Nicholas R Perl et al.
Organic letters, 9(18), 3699-3701 (2007-08-10)
A new chiral allylchlorosilane has been developed that allows the highly enantioselective allylation and crotylation of a range of 2-imidazolylaldimines and ketimines. The method may be exploited for the protecting group-free synthesis of a diverse array of imidazole-bearing chiral carbinamines

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service