Skip to Content
Merck
All Photos(1)

Documents

240958

Sigma-Aldrich

Propylamine

≥99%

Synonym(s):

1-Aminopropane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2NH2
CAS Number:
Molecular Weight:
59.11
Beilstein:
1098243
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39030103
PubChem Substance ID:
NACRES:
NA.22

vapor density

2 (vs air)

vapor pressure

4.79 psi ( 20 °C)

Assay

≥99%

form

liquid

autoignition temp.

604 °F

expl. lim.

10.4 %

refractive index

n20/D 1.388 (lit.)

bp

48 °C (lit.)

mp

−83 °C (lit.)

solubility

water: soluble 0.4 part(lit.)
chloroform: soluble 1.5 part(lit.)

density

0.719 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCN

InChI

1S/C3H9N/c1-2-3-4/h2-4H2,1H3

InChI key

WGYKZJWCGVVSQN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Propylamine is a primary amine and an important building block in the chemical synthesis. It is generally used as a petroleum additive and solvent in organic syntheses. It can also be used as a finishing agent for paints, drugs, rubber, pesticides, textile, and resin. It is used in the production of fungicides, antivirals, catalysts, molecular sieves, and corrosion inhibitors.

Application

Propylamine was used in the synthesis of dendritic polymer-modified silica (DPS) stationary phase by consecutive amine-epoxy reactions with aniline and 1,4-butanedioldiglycidyl ether.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

<-31.0 °F

Flash Point(C)

< -35 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Unimolecular decomposition reactions of propylamine and protonated propylamine
Almatarneh MH, et al.
ACS Omega, 4(2), 3306-3313 (2019)
Pawel Misiak et al.
International journal of nanomedicine, 15, 7263-7278 (2020-10-17)
Efficient intracellular delivery of a therapeutic compound is an important feature of smart drug delivery systems (SDDS). Modification of a carrier structure with a cell-penetrating ligand, ie, cholesterol moiety, is a strategy to improve cellular uptake. Cholesterol end-capped poly(N-isopropylacrylamide)s offer
Timmy Mani et al.
ChemMedChem, 8(12), 1963-1977 (2013-10-12)
The urokinase receptor (uPAR) is a cell-surface protein that is part of an intricate web of transient and tight protein interactions that promote cancer cell invasion and metastasis. Here, we evaluate the binding and biological activity of a new class
Marian Cors et al.
Scientific reports, 9(1), 13812-13812 (2019-09-27)
The peculiar linear temperature-dependent swelling of core-shell microgels has been conjectured to be linked to the core-shell architecture combining materials of different transition temperatures. Here the structure of pNIPMAM-core and pNNPAM-shell microgels in water is studied as a function of
Yun Li et al.
Journal of chromatography. A, 1337, 133-139 (2014-03-19)
A novel dendritic polymer-modified silica (DPS) stationary phase was prepared by a divergent synthesis scheme starting from propylamine on silica by consecutive amine-epoxy reactions with 1,4-butanedioldiglycidyl ether and aniline. Both elemental analysis and infrared spectra data shows the successful growth

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service