Skip to Content
Merck
All Photos(4)

Documents

471208

Sigma-Aldrich

Ethylamine solution

66.0-72.0% in H2O

Synonym(s):

Aminoethane, Monoethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5NH2
CAS Number:
Molecular Weight:
45.08
Beilstein:
505933
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

concentration

66.0-72.0% in H2O

density

0.81 g/mL at 20 °C

SMILES string

CCN

InChI

1S/C2H7N/c1-2-3/h2-3H2,1H3

InChI key

QUSNBJAOOMFDIB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethylamine solution is used in the preparation of N-ethyl triazineepiperazine copolymer (ETPC), as a hydrophobic flame retardant material. It is also used in the modification of poly(tetramethylene oxide)/silica hybrid composites.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

71.6 °F

Flash Point(C)

22 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Modification of an inorganic-organic hybrid composite: effect of ethylamine.
Brennan, Anthony B and Miller, Thomas M
Chemistry of Materials, 6(3), 262-267 (1994)
Synthesis of N-ethyl triazine-piperazine copolymer and flame retardancy and water resistance of intumescent flame retardant polypropylene.
Yang, Kun et al.
Polymer Degradation and Stability, 98(7), 1397-1406 (2013)
Paige E Mahaney et al.
Journal of medicinal chemistry, 51(13), 4038-4049 (2008-06-19)
Further exploration of the cycloalkanol ethylamine scaffold, of which venlafaxine ( 1) is a member, was undertaken to develop novel and selective norepinephrine reuptake inhibitors (NRIs) for evaluation in a variety of predictive animal models. These efforts led to the
Stacey F Bent et al.
Proceedings of the National Academy of Sciences of the United States of America, 108(3), 956-960 (2010-11-12)
Surface functionalization of semiconductors has been the backbone of the newest developments in microelectronics, energy conversion, sensing device design, and many other fields of science and technology. Over a decade ago, the notion of viewing the surface itself as a
Riccardo Di Corato et al.
ACS nano, 5(2), 1109-1121 (2011-01-12)
Trifunctional polymer nanobeads are prepared by destabilization of a mixture of magnetic nanoparticles, quantum dots, and an amphiphilic polymer, followed by functionalization of the bead surface with folic acid molecules. The distribution of the nanoparticles within the nanobeads can be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service