235555
6-Bromohexanoyl chloride
97%
Synonym(s):
6-Bromocaproyl chloride, 6-Bromohexanoic acid chloride
About This Item
Recommended Products
Assay
97%
form
liquid
refractive index
n20/D 1.486 (lit.)
bp
130 °C/20 mmHg (lit.)
density
1.395 g/mL at 25 °C (lit.)
functional group
acyl chloride
SMILES string
ClC(=O)CCCCCBr
InChI
1S/C6H10BrClO/c7-5-3-1-2-4-6(8)9/h1-5H2
InChI key
HBPVGJGBRWIVSX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- (S)- or (R)-N-(6-bromohexanoyl)-α-methylbenzylamine
- 1-bromoheptadecan-6-one and 1-bromooctadecan-6-one
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B - STOT SE 3
Target Organs
Respiratory system
Supplementary Hazards
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
230.0 °F - closed cup
Flash Point(C)
110 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service