Skip to Content
Merck
All Photos(1)

Key Documents

251933

Sigma-Aldrich

4-Bromobutyryl chloride

95%

Synonym(s):

4-Bromobutanoic acid chloride, 4-Bromobutanoyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2CH2CH2COCl
CAS Number:
Molecular Weight:
185.45
Beilstein:
1743026
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

reaction suitability

reagent type: cross-linking reagent

refractive index

n20/D 1.492 (lit.)

bp

101 °C/37 mmHg (lit.)

density

1.602 g/mL at 25 °C (lit.)

functional group

bromo

SMILES string

BrCCCC(Cl)=O

InChI

1S/C4H6BrClO/c5-3-1-2-4(6)7/h1-3H2

InChI key

LRTRXDSAJLSRTG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Pharmaceutical research applications: 4-Bromobutyryl chloride was utilized in the synthesis of newly developed 3,4,5-trisubstituted-1,2,4-triazole derivatives. These derivatives demonstrated significant alpha-glucosidase inhibitory potential, suggesting their utility in managing diabetes through inhibition of carbohydrate hydrolyzing enzymes. This application underscores the importance of 4-Bromobutyryl chloride in the development of bioactive molecules for potential therapeutic use (Nafeesa et al., 2019).

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service