Skip to Content
Merck
All Photos(2)

Key Documents

142514

Sigma-Aldrich

3-Bromopropionyl chloride

technical grade

Synonym(s):

β-Bromopropanoyl chloride, β-Bromopropionyl chloride, ψ-Bromopropanoyl chloride, 3-Bromopropanoic acid chloride, 3-Bromopropanoyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2CH2COCl
CAS Number:
Molecular Weight:
171.42
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

form

liquid

refractive index

n20/D 1.49 (lit.)

bp

55-57 °C/17 mmHg (lit.)

density

1.701 g/mL at 25 °C (lit.)

functional group

acyl chloride
bromo

storage temp.

2-8°C

SMILES string

ClC(=O)CCBr

InChI

1S/C3H4BrClO/c4-2-1-3(5)6/h1-2H2

InChI key

IHBVNSPHKMCPST-UHFFFAOYSA-N

General description

3-Bromopropionyl chloride reacts with poly(ethylene glycol) methacrylate to yield brominated poly(ethylene glycol) methacrylate. It causes the acylation of 4-aryl-substituted 3,4-di-hydropyrimidine(1H)-2-thiones to give bicyclic pyrimido[2,1-b][1,3]thiazines.

Application

3-Bromopropionyl chloride was used in the preparation of 1-chloro-4-bromo-2-butanone.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and Characterization of a Novel Macromonomer Initiator for Reversible Addition Fragmentation Chain Transfer (RAFT). Evaluation of the Polymerization Kinetics and Gelation Behaviors.
Ozturk T and Hazer B.
Journal of Macromolecular Science, Part A: Pure and Applied Chemistry, 47(3), 265-272 (2010)
Synthesis of bicyclic pyrimido [2, 1-b][1, 3] thiazines based on 3, 4-dihydropyrimidine-(1H)-2-thiones.
Kulakov IV and Turdybekov DM.
Chemistry of Heterocyclic Compounds, 46(3), 342-346 (2010)
A Nagy et al.
Proceedings of the National Academy of Sciences of the United States of America, 93(6), 2464-2469 (1996-03-19)
A convenient, high yield conversion of doxorubicin to 3'-deamino-3'-(2''-pyrroline-1''-yl)doxorubicin is described. This daunosamine-modified analog of doxorubicin is 500-1000 times more active in vitro than doxorubicin. The conversion is effected by using a 30-fold excess of 4-iodobutyraldehyde in anhydrous dimethylformamide. The
Ji Eon Chae et al.
Polymers, 13(5) (2021-03-07)
Polystyrene-based polymers with variable molecular weights are prepared by radical polymerization of styrene. Polystyrene is grafted with bromo-alkyl chains of different lengths through Friedel-Crafts acylation and quaternized to afford a series of hydroxide-ion-conducting ionomers for the catalyst binder for the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service