Skip to Content
Merck
All Photos(2)

Key Documents

8.08260

Sigma-Aldrich

Trifluoroacetic acid

for synthesis

Synonym(s):

TFA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3COOH
CAS Number:
Molecular Weight:
114.02
Beilstein:
742035
MDL number:
UNSPSC Code:
12352106
EC Index Number:
200-929-3
NACRES:
NA.22
Assay:
≥99% (acidimetric)
bp:
72.4 °C (lit.)
vapor pressure:
97.5 mmHg ( 20 °C)
Pricing and availability is not currently available.

vapor density

3.9 (vs air)

Quality Level

vapor pressure

97.5 mmHg ( 20 °C)

Assay

≥99% (acidimetric)

form

liquid

refractive index

n20/D 1.3 (lit.)

pH

1 (10 g/L in H2O)

bp

72.4 °C (lit.)

mp

−15.4 °C (lit.)

solubility

soluble 10 g/mL

density

1.489 g/mL at 20 °C (lit.)

storage temp.

2-30°C

SMILES string

OC(C(F)(F)F)=O

InChI

1S/C2HF3O2/c3-2(4,5)1(6)7/h(H,6,7)

InChI key

DTQVDTLACAAQTR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trifluoroacetic acid is widely used in organic synthesis as a solvent, catalyst and reagent. It is a strong acid and low boiling point reagent. It can be used as a catalyst for a variety of reactions such as rearrangements, functional group deprotections, oxidations, reductions, condensations, hydroarylations and trifluoromethylations.[1]

Application

Trifluoroacetic acid can be used as a catalyst

  • In the direct conversion of cyclohexanone to caprolactam using acetonitrile as the additive.
  • In the synthesis of 1,2,4,5-tetrasubstituted imidazoles by four-component condensation of benzil, aldehydes, amines, and ammonium acetate.
Trifluoroacetic acid can also be used
  • In the synthesis of d-α-Tterpineo from d-limonene by Markovnikov addition followed by hydrolysis.

Features and Benefits

Trifluoroacetic acid can be used as a catalyst due to its
  • High acidity
  • Easy elimination
  • Good solubility in organic solvent and in water.

Analysis Note

Assay (acidimetric): ≥ 99.0 %
Density (d 20 °C/ 4 °C): 1.487 - 1.490
Identity (IR): passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

>212.0 °F - Pensky-Martens closed cup

Flash Point(C)

> 100 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

A Practical Synthesis of?d-?-Terpineol via Markovnikov Addition of?d-Limonene Using Trifluoroacetic Acid
Yuasa Y, et al.
Organic Process Research & Development, 10(6), 1231-1232 (2006)
Trifluoroacetic acid: Uses and recent applications in organic synthesis
Lopez SE, et al.
Journal of Fluorine Chemistry, 156, 73-100 (2013)
Trifluoroacetic acid as an efficient catalyst for one-pot, four-component synthesis of 1, 2, 4, 5-tetrasubstituted imidazoles under microwave-assisted, solvent-free conditions
Mohammadizadeh MR, et al.
Synthetic Communications, 39(18), 3232-3242 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service