Skip to Content
Merck
All Photos(2)

Key Documents

226505

Sigma-Aldrich

2,2,2-Trichloro-1,1-dimethylethyl chloroformate

96%

Synonym(s):

β,β,β-Trichloro-tert-butyl chloroformate, β,β,β-Trichloro-tert-butoxycarbonyl chloride, TCBoc-chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCO2C(CH3)2CCl3
CAS Number:
Molecular Weight:
239.91
Beilstein:
1935464
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

solid

bp

83-84 °C/14 mmHg (lit.)

mp

28-30 °C (lit.)

solubility

ethanol: soluble 10%, clear, colorless

storage temp.

2-8°C

SMILES string

CC(C)(OC(Cl)=O)C(Cl)(Cl)Cl

InChI

1S/C5H6Cl4O2/c1-4(2,5(7,8)9)11-3(6)10/h1-2H3

InChI key

GMELMFSDPDSXOZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The solvolysis rate constants of 2,2,2-trichloro-1,1-dimethylethyl chloroformate in binary and pure solvents was studied.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kinetic studies of the solvolyses of 2, 2, 2-trichloro-1, 1-dimethylethyl chloroformate.
Bull. Korean Chem. Soc., 31(4), 835-835 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service