Skip to Content
Merck
All Photos(2)

Key Documents

142077

Sigma-Aldrich

2,2,2-Trichloroethyl chloroformate

98%

Synonym(s):

2,2,2-Trichloroethoxycarbonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCOOCH2CCl3
CAS Number:
Molecular Weight:
211.86
Beilstein:
970619
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.06 psi ( 20 °C)

Assay

98%

form

liquid

refractive index

n20/D 1.471 (lit.)

bp

171-172 °C (lit.)

density

1.539 g/mL at 25 °C (lit.)

functional group

chloro

storage temp.

2-8°C

SMILES string

ClC(=O)OCC(Cl)(Cl)Cl

InChI

1S/C3H2Cl4O2/c4-2(8)9-1-3(5,6)7/h1H2

InChI key

LJCZNYWLQZZIOS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,2,2-Trichloroethyl chloroformate (TrocCl) serves as a versatile reagent used for selective acylation and dealkylation in organic synthesis. It is used in regio, chemo, and stereoselective synthesis.

Application

  • 2,2,2-Trichloroethyl chloroformate was used as derivatizing reagent in gas chromatographic/mass spectrometric determination of a large range of amphetamine-related drugs and ephedrines in plasma, urine and hair samples.
  • It was used as starting reagent during the synthesis of 6-Nor-9,10-dihydrolysergic acid methyl ester.
  • It was used as reagent during N-demethylation of dextromethorphan.
  • It was used as protecting reagent for aliphatic and aromatic hydroxyl and amino groups.
Protecting reagent for aliphatic and aromatic hydroxyl and amino groups.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giampietro Frison et al.
Rapid communications in mass spectrometry : RCM, 19(7), 919-927 (2005-03-05)
A new analytical approach, based on derivatization with 2,2,2-trichloroethyl chloroformate and gas chromatography/mass spectrometry (GC/MS), was investigated for qualitative and quantitative analyses of a large range of amphetamine-related drugs and ephedrines in plasma, urine and hair samples. Sample preparation involved
J O Kuye et al.
Journal of chromatography, 272(2), 307-313 (1983-02-11)
A specific method for the gas chromatographic determination of chloroquine (CQ) after derivatization with chloroformates, using 9-bromophenanthrene as the internal standard and a column filled with 3% OV-17 on 80-100 mesh Supelcoport is described. Derivatization with chloroformates produced a pyrrolidine
Gas chromatography/mass spectrometry determination of mephedrone in drug seizures after derivatization with 2,2,2-trichloroethyl chloroformate.
Giampietro Frison et al.
Rapid communications in mass spectrometry : RCM, 25(2), 387-390 (2010-12-31)
A Dasgupta et al.
American journal of clinical pathology, 109(5), 527-532 (1998-05-12)
Amphetamine and methamphetamine are commonly abused central nervous system stimulants. We describe a rapid new derivatization of amphetamine and methamphetamine using 2,2,2-trichloroethyl chloroformate for gas chromatography-mass spectrometric analysis. Amphetamine and methamphetamine, along with N-propyl amphetamine (internal standard), were extracted from
A M Crider et al.
Journal of pharmaceutical sciences, 70(12), 1319-1321 (1981-12-01)
6-Nor-9,10-dihydrolysergic acid methyl ester (IV) was prepared by demethylation of 9,10-dihydrolysergic acid methyl ester (II) with 2,2,2-trichloroethyl chloroformate, followed by reduction of the intermediate carbamate (III) with zinc in acetic acid. The 6-ethyl-V and 6-n-propyl-VI derivatives were prepared by alkylation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service