Skip to Content
Merck
All Photos(1)

Key Documents

185892

Sigma-Aldrich

Ethyl chloroformate

97%

Synonym(s):

Chloroformic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCOOC2H5
CAS Number:
Molecular Weight:
108.52
Beilstein:
385653
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.74 (vs air)

Quality Level

vapor pressure

3.42 psi ( 20 °C)

Assay

97%

form

liquid

autoignition temp.

842 °F

refractive index

n20/D 1.395 (lit.)

bp

93 °C (lit.)

mp

−81 °C (lit.)

solubility

alcohol: miscible
benzene: miscible
chloroform: miscible
diethyl ether: miscible
water: insoluble

density

1.135 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

CCOC(Cl)=O

InChI

1S/C3H5ClO2/c1-2-6-3(4)5/h2H2,1H3

InChI key

RIFGWPKJUGCATF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

  • Ethyl chloroformate was used in the synthesis of nitrile oxides.
  • It was used as derivatization reagent to investigate stereochemical conversion of chiral non-steroidal anti-inflammatory drugs during derivatization reaction.
  • It was used to develop flexible template strategy for the generation of nanometer-scale templates via dip-pen nanolithography.
  • It was used as derivatization reagent in an enantiomeric analysis of amino acids by two-dimensional gas chromatography.
  • It was also used with ammonia and cyanuric chloride to convert carboxylic acids to nitriles.

Packaging

Derivatization reagent employed in an enantiomeric analysis of amino acids by two-dimensional gas chromatography. Also used with ammonia and cyanuric chloride to convert carboxylic acids to nitriles.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

60.8 °F

Flash Point(C)

16 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sezin Erarpat et al.
Journal of separation science, 43(6), 1100-1106 (2019-12-21)
A green and fast analytical method for the determination of l-methionine in human plasma is presented in this study. Preconcentration of the analyte was carried out by switchable solvent liquid phase microextraction after ethyl chloroformate derivatization reaction. Instrumental detection of
A convenient preparative method of nitrile oxides by the dehydration of primary nitro compounds with ethyl chloroformate or benzenesulfonyl chloride in the presence of triethylamine.
Shimizu T, et al.
Bulletin of the Chemical Society of Japan, 59(9), 2827-2831 (1986)
Melanie Junge et al.
Chirality, 19(3), 228-234 (2007-01-19)
The chiral separation of amino acids (AA) derivatised with ethyl chloroformate by using comprehensive two-dimensional gas chromatography is reported. A commercially available enantioselective capillary column (Chirasil-l-Val) has been tested as first-dimension column. Two nonenantioselective stationary phases (BPX50 and BP1) with
M R Wright et al.
Journal of chromatography, 616(1), 59-65 (1993-06-23)
A potential problem with chiral derivatization is the possibility of stereochemical conversion during the derivatization reaction. This possibility has been examined using the non-steroidal anti-inflammatory drugs ibuprofen, ketoprofen, etodolac and flurbiprofen. To avoid possible interference from stereochemical impurities, male Sprague-Dawley
Mohana Krishna Reddy Mudiam et al.
Analytical and bioanalytical chemistry, 401(5), 1695-1701 (2011-07-12)
A simple and rapid analytical method based on in-matrix ethyl chloroformate (ECF) derivatization has been developed for the quantitative determination of bisphenol-A (BPA) in milk and water samples. The samples containing BPA were derivatised with ECF in the presence of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service