225959
Magnesium bromide ethyl etherate
99%
Synonym(s):
Dibromo(1,1′-oxybis(ethane))magnesium, Dibromo(diethyl ether)magnesium, Magnesium bromide diethyl etherate, Magnesium bromide etherate
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
MgBr2 · O(C2H5)2
CAS Number:
Molecular Weight:
258.23
Beilstein:
4937320
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
99%
functional group
ether
SMILES string
Br[Mg]Br.CCOCC
InChI
1S/C4H10O.2BrH.Mg/c1-3-5-4-2;;;/h3-4H2,1-2H3;2*1H;/q;;;+2/p-2
InChI key
JGZKUKYUQJUUNE-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Magnesium bromide ethyl etherate can be used as a reagent:
- In the Lewis acid-catalyzed Diels-Alder reactions.
- For the conversion of aromatic aldehydes into alcohols and carboxylic acids via Cannizzaro reaction.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Sol. 1 - Water-react 2
Storage Class Code
4.3 - Hazardous materials which set free flammable gases upon contact with water
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Room-temperature Cannizzaro reaction under mild conditions facilitated by magnesium bromide ethyl etherate and triethylamine
Abaee M, et al.
Organic Letters, 7, 5893-5895 (2005)
Tetrahedron Asymmetry, 2, 1193-1193 (1991)
Synthesis and Diels-Alder reactions of homochiral 2-sulfinylmaleates with cyclopentadiene
Alonso I, et al.
Tetrahedron Asymmetry, 2, 1193-1207 (1991)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service