Skip to Content
Merck
All Photos(3)

Key Documents

683396

Sigma-Aldrich

[Pd(allyl)Cl]2

Umicore

Synonym(s):

Allylpalladium(II) chloride dimer, Allylchloropalladium dimer, Bis(η3-allyl)di(μ-chloro)dipalladium(II), Bis(allyl)dichlorodipalladium, Bis(allyl)dichloropalladium, Bis(allylchloropalladium), Diallyldichlorodipalladium, [PdCl(C3H5)]2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10Cl2Pd2
CAS Number:
Molecular Weight:
365.89
Beilstein:
4124623
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (C, elemental analysis)

form

solid

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

storage temp.

2-8°C

SMILES string

Cl[Pd]CC=C.Cl[Pd]CC=C

InChI

1S/2C3H5.2ClH.2Pd/c2*1-3-2;;;;/h2*3H,1-2H2;2*1H;;/q;;;;2*+1/p-2

InChI key

TWKVUTXHANJYGH-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

[Pd(allyl)Cl]2 can be used as a catalyst:
  • For the silylation of organobromides.
  • To synthesize α-aryl carbonyl compounds by coupling reaction between aldehydes and aryl halides.
  • To prepare various arylthiophene derivatives by cross-coupling reaction with aryl halides via C-H functionalization.

Legal Information

Product of Umicore

Additional information available at www.pmc.umicore.com

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synthesis of HOMSi Reagents by Pd/Cu-Catalyzed Silylation of Bromoarenes with Disilanes
Minami Y, et al.
Chemistry Letters (Jpn), 43(2), 201-203 (2014)
Direct Arylation of Thiophenes via Palladium-Catalysed C-H Functionalisation at Low Catalyst Loadings
Battace A, et al.
Advanced Synthesis & Catalysis, 349(16), 2507-2516 (2007)
Direct Arylation of Thiophenes via Palladium-Catalysed C-H Functionalisation at Low Catalyst Loadings
Battace A, et al.
advanced synthesis and catalysis, 349(16), 2507-2516 (2007)
Palladium-catalyzed alpha-arylation of aldehydes with bromo- and chloroarenes catalyzed by [{Pd(allyl)Cl}2] and dppf or Q-phos
Giang D Vo and John F Hartwig
Angewandte Chemie (International ed. in English), 2127-2130 (2008)
Palladium-catalyzed alpha-arylation of aldehydes with bromo- and chloroarenes catalyzed by [{Pd(allyl)Cl}2] and dppf or Q-phos
Giang D Vo and John F Hartwig
Angewandte Chemie (International Edition in English), 2127-2130 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service