Skip to Content
Merck
All Photos(2)

Key Documents

230170

Sigma-Aldrich

Tetrabutylammonium borohydride

98%

Synonym(s):

Tetra-n-butylammonium tetrahydridoborate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(BH4)
CAS Number:
Molecular Weight:
257.31
Beilstein:
4212332
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

reaction suitability

reagent type: reductant

mp

124-128 °C (lit.)

functional group

amine

SMILES string

[BH4-].CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.BH4/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;/h5-16H2,1-4H3;1H4/q+1;-1

InChI key

GMBOFJFPOCGSOI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetrabutylammonium borohydride is used for the reduction of carboxylic acids, aldehydes, and ketones.

Application

Selective reducing agent for β-ketoamides, β-ketoesters, and carbohydrate derivatives.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - Water-react 2

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrabutylammonium borohydride
Raber DJ and Guida WC
eEROS (Encyclopedia of Reagents for Organic Synthesis) (2001)
Chemistry Letters (Jpn), 1469-1469 (1992)
Tetrahedron, 49, 11169-11169 (1993)
Matthew N McCarroll et al.
Nature communications, 10(1), 4078-4078 (2019-09-11)
Anesthetics are generally associated with sedation, but some anesthetics can also increase brain and motor activity-a phenomenon known as paradoxical excitation. Previous studies have identified GABAA receptors as the primary targets of most anesthetic drugs, but how these compounds produce
Li Zhou et al.
Nanoscale, 12(3), 1484-1494 (2020-01-08)
Artificial limbs have been widely investigated in the past several decades, and multifuncional bionic limbs have already been constructed. However, due the lack of nociceptive systems, amputees still cannot feel ubiquitous noxious stimuli through bionic limbs. The construction of artificial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service