Skip to Content
Merck
All Photos(1)

Key Documents

223182

Sigma-Aldrich

α-Terpinene

85%

Synonym(s):

alpha-Terpinene, 1-Isopropyl-4-methyl-1,3-cyclohexadiene, p-Mentha-1,3-diene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
Beilstein:
1853379
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

85%

form

liquid

refractive index

n20/D 1.478 (lit.)

bp

173-175 °C (lit.)

solubility

ethanol: soluble 100 mg/mL, clear, colorless

density

0.837 g/mL at 25 °C (lit.)

SMILES string

CC(C)C1=CC=C(C)CC1

InChI

1S/C10H16/c1-8(2)10-6-4-9(3)5-7-10/h4,6,8H,5,7H2,1-3H3

InChI key

YHQGMYUVUMAZJR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

α-Terpinene, a volatile essential oil derived from Melaleuca alternifolia, shows antimicrobial properties against various human pathogens.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Sens. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

122.0 °F - closed cup

Flash Point(C)

50 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tais Susane Pereira et al.
Journal of ethnopharmacology, 151(2), 852-857 (2013-12-10)
The volatile essential oil derived from the plant Melaleuca alternifolia, also called tea tree oil (TTO), is largely employed for its antimicrobial properties against several human pathogens. It is used in many topical formulations to treat cutaneous infections. Since very
D Djenane et al.
Food science and technology international = Ciencia y tecnologia de los alimentos internacional, 17(6), 505-515 (2011-11-04)
Essential oils (EOs) extracted by hydrodistillation from leaf parts of Algerian Eucalyptus globulus, Myrtus communis and Satureja hortensis were analyzed by gas chromatography/mass spectrometry (GC/MS). The main components of EOs obtained were γ-terpinene (94.48%), 1,8-cineole (46.98%) and carvacrol (46.10%), respectively
Mohammad S Abu-Darwish et al.
Natural product research, 26(14), 1310-1317 (2011-06-29)
The effect of plant space and time of harvesting on yield and quality of Thymus vulgaris was evaluated in Jordan. Thyme was cultivated in rows of 50 cm apart with inter-row spacing of 15, 30 or 45 cm and was grown at
Ekhteraei Tousi Samaneh et al.
Natural product research, 24(19), 1834-1842 (2010-11-26)
Essential oils from the subterranean organs of three species of Valeriana L. from Iran (Valeriana sisymbriifolia Vahl, Valeriana alliariifolia Adams and Valeriana officinalis L.) belonging to Valerianaceae family have been obtained by hydrodistillation and analysed by gas chromatography-mass spectrometry in
Nataraj Jagannath et al.
Natural product communications, 7(7), 943-946 (2012-08-23)
The essential oil was extracted from the seeds of Heracleum rigens by hydrodistillation and a total of twenty compounds accounting for 98.5% of the total oil composition were identified. Physicochemical properties and chemical composition of the oil was determined by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service