Skip to Content
Merck
All Photos(1)

Documents

C100005

Sigma-Aldrich

1,3-Cyclohexadiene

contains 0.05% BHT as inhibitor, 97%

Synonym(s):

1,2-Dihydrobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8
CAS Number:
Molecular Weight:
80.13
Beilstein:
506024
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

contains

0.05% BHT as inhibitor

refractive index

n20/D 1.474 (lit.)

bp

80 °C (lit.)

density

0.841 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C1CC=CC=C1

InChI

1S/C6H8/c1-2-4-6-5-3-1/h1-4H,5-6H2

InChI key

MGNZXYYWBUKAII-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,3-Cyclohexadiene can undergo:
  • C-C coupling with aromatic alcohols via iridium-catalyzed hydrogen auto-transfer and with aldehydes via transfer hydrogenation mediated by isopropanol to form carbonyl addition products.
  • Living anionic polymerization with n-BuLi/TMEDA system to form polycyclohexadiene.
  • Platinum-catalyzed silaboration to form (1R,4S)-1-(dimethylphenylsilyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-cyclohexene.
  • Aerobic palladium-catalyzed 1,4-diacetoxylation in the presence of cobalt tetra(hydroquinone)porphyrin as an electron transfer reagent.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

66.0 °F

Flash Point(C)

18.9 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew D Cohen et al.
Journal of the American Chemical Society, 125(6), 1444-1445 (2003-02-06)
Benzoyl nitroside (5) was generated in solution by laser photolysis of 3,5-diphenyl-1,2,4-oxadiazole-4-oxide (4) and studied by time-resolved infrared spectroscopy. The second-order rate constants for reaction of 5 with diethylamine and 1,3-cyclohexadiene were determined to be (1.3 +/- 0.5) x 10(5)
Sarah J Ryan et al.
Journal of the American Chemical Society, 133(13), 4694-4697 (2011-03-12)
Herein we report the first all-carbon N-heterocyclic carbene-catalyzed (4 + 2) cycloaddition. The reaction proceeds with α,β-unsaturated acid fluorides and silyl dienol ethers and produces 1,3-cyclohexadienes with complete diastereocontrol (dr >20:1) while demonstrating a new type of reaction cascade exploiting
Xiang-Shan Wang et al.
Journal of combinatorial chemistry, 11(6), 1011-1022 (2009-09-22)
An efficient and green method for the synthesis of highly substituted cyclohexa-1,3-diene, polyhydroindene, polyhydronaphthalene, isochromene, isothiochromene, and isoquinoline derivatives has been developed. The synthesis was achieved by using a multicomponent procedure in ionic liquid media. The features of this procedure
John F Bower et al.
Organic letters, 10(5), 1033-1035 (2008-02-08)
Under hydrogen autotransfer conditions employing a catalyst derived from [Ir(cod)Cl]2 and BIPHEP, 1,3-cyclohexadiene (CHD) couples to benzylic alcohols 1a-9a to furnish carbonyl addition products 1c-9c, which appear as single diastereomers with variable quantities of regioisomeric adducts 1d-9d. Under related transfer
Borislav Naydenov et al.
Nano letters, 6(8), 1752-1756 (2006-08-10)
We present a study that maps out chemical bond formation between a Pt-inked probe and a single 1,3-cyclohexadiene (1,3-CHD) molecule on Si(100). By separating the mechanical and electronic contributions to the current during the approach to contact, we show that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service