Skip to Content
Merck
All Photos(1)

Key Documents

W235601

Sigma-Aldrich

p-Cymene

≥97%, FG

Synonym(s):

1-Isopropyl-4-methylbenzene, 4-Isopropyltoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4CH(CH3)2
CAS Number:
Molecular Weight:
134.22
FEMA Number:
2356
Beilstein:
1903377
EC Number:
Council of Europe no.:
620
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
1.002
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

vapor density

4.62 (vs air)

vapor pressure

1.5 mmHg ( 20 °C)
3.7 mmHg ( 37.7 °C)

Assay

≥97%

autoignition temp.

817 °F

expl. lim.

5.6 %

refractive index

n20/D 1.490 (lit.)

bp

176-178 °C (lit.)

density

0.86 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

spicy; woody; citrus

SMILES string

CC(C)c1ccc(C)cc1

InChI

1S/C10H14/c1-8(2)10-6-4-9(3)5-7-10/h4-8H,1-3H3

InChI key

HFPZCAJZSCWRBC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 3 - Repr. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

125.6 °F - closed cup

Flash Point(C)

52 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gerd Ludwig et al.
Journal of inorganic biochemistry, 113, 77-82 (2012-06-16)
Reactions of ω-diphenylphosphino-functionalized alkyl phenyl sulfides Ph(2)P(CH(2))(n)SPh (n=1, L1; 2, L2; 3, L3), sulfoxides Ph(2)P(CH(2))(n)S(O)Ph (n=1, L4; 2, L5; 3, L6) and sulfones Ph(2)P(CH(2))(n)S(O)(2)Ph (n=1, L7; 2, L8; 3, L9) with the dinuclear chlorido bridged ruthenium(II) complex [{Ru(η(6)-p-cymene)Cl(2)}(2)] afforded mononuclear
Rajenahally V Jagadeesh et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(51), 14375-14379 (2011-11-25)
An easily accessible in situ catalyst composed of [{RuCl(2)(p-cymene)}(2)] and terpyridine has been developed for the selective transfer hydrogenation of aromatic nitro and azo compounds. The procedure is general and the selectivity of the catalyst has been demonstrated by applying
Marek Kuzma et al.
Dalton transactions (Cambridge, England : 2003), 42(14), 5174-5182 (2013-02-14)
Asymmetric transfer hydrogenation (ATH) of cyclic imines using [RuCl(η(6)-p-cymene)TsDPEN] (TsDPEN = N-tosyl-1,2-diphenylethylenediamine) was tested with various aliphatic (secondary, tertiary) and aromatic amines employed in the HCOOH-base hydrogen donor mixture. Significant differences in reaction rates and stereoselectivity were observed, which pointed
Nehad E M Taktak et al.
Pesticide biochemistry and physiology, 158, 156-165 (2019-08-06)
Culex pipiens is a main vector for Bancroftian filariasis, Rift Valley Fever and diseases caused by other viruses, leaving several peoples with disabilities. In recent years, plant derived compounds have received much attention as potential alternatives to synthetic chemicals due
Lutz Ackermann et al.
Organic letters, 14(8), 2146-2149 (2012-04-13)
The ruthenium(II) carboxylate complex [Ru(O(2)CMes)(2)(p-cymene)] enabled efficient direct arylations of unactivated C-H bonds with easily available, inexpensive phenols. Extraordinary chemoselectivity of the well-defined ruthenium catalyst set the stage for challenging C-H/C-O bond functionalizations to occur under solvent-free conditions as well

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service