Skip to Content
Merck
All Photos(3)

Key Documents

W285611

Sigma-Aldrich

α-Phellandrene

greener alternative

natural, ≥85%, FG

Synonym(s):

p-Mentha-1,5-diene, Menthadiene, 2-Methyl-5-(1-methylethyl)-1,3-cyclohexadiene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
FEMA Number:
2856
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
1.006
NACRES:
NA.21

grade

FG
Halal
Kosher
natural

Quality Level

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

Assay

≥85%

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.474 (lit.)

density

0.85 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

greener alternative category

Organoleptic

spicy; woody

SMILES string

CC(C)C1CC=C(C)C=C1

InChI

1S/C10H16/c1-8(2)10-6-4-9(3)5-7-10/h4-6,8,10H,7H2,1-3H3

InChI key

OGLDWXZKYODSOB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

α-Phellandrene is monoterpene reported to be one of the aroma compounds of black (Piper nigrum) and white ‘‘Ashanti pepper′′ (Piper guineense). It also occurs in the leaves of basil.
We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”.

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

116.6 °F

Flash Point(C)

47 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aroma compound analysis of Piper nigrum and Piper guineense essential oils from Cameroon using solid-phase microextraction?gas chromatography, solid-phase microextraction?gas chromatography?mass spectrometry and olfactometry.
Jirovetz L, et al.
Journal of Chromatography A, 976(1), 265-275 (2002)
Comparative essential oil composition of flowers, leavesand stems of basil (Ocimum basilicum L.) used as herb.
Chalchat J C & Ozcan MM.
Food Chemistry, 110(2), 501-503 (2008)
Joachim Morrens et al.
Neuron, 106(1), 142-153 (2020-02-07)
Dopamine neurons mediate the association of conditioned stimuli (CS) with reward (unconditioned stimuli, US) by signaling the discrepancy between predicted and actual reward during the US. Some theoretical models suggest that learning is also influenced by the salience or associability

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service