Skip to Content
Merck
All Photos(1)

Key Documents

W424201

Sigma-Aldrich

Amylamine

≥99%, FG

Synonym(s):

Pentylamine, 1-Aminopentane, n-Amylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4NH2
CAS Number:
Molecular Weight:
87.16
FEMA Number:
4242
Beilstein:
505953
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
11.021
NACRES:
NA.21
grade:
FG
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR part 172.515

vapor density

3.01 (vs air)

Assay

≥99%

expl. lim.

22 %

refractive index

n20/D 1.411 (lit.)

bp

104 °C (lit.)

mp

−50 °C (lit.)

density

0.752 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fishy

SMILES string

CCCCCN

InChI

1S/C5H13N/c1-2-3-4-5-6/h2-6H2,1H3

InChI key

DPBLXKKOBLCELK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

44.6 °F

Flash Point(C)

7 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Widy-Tyszkiewicz et al.
Polish journal of pharmacology and pharmacy, 35(6), 467-472 (1983-11-01)
The effect of butylamine, pentylamine and hexylamine on spontaneous locomotor activity of mice was evaluated in the presence or absence of drugs affecting monoaminergic neurons. Pentylamine and hexylamine produced similar effects (stimulation after low, inhibition after high doses), while butylamine
E Widy-Tyszkiewicz et al.
Pharmacology, biochemistry, and behavior, 13(3), 385-390 (1980-09-01)
Motor and autonomic effects of n-pentylamine in mice have been evaluated in relation to aminergic mechanisms by combining administration of the amine with drugs having well known effects on such mechanisms. Some evidence has been found to indicate the importance
Shawn P Vorce et al.
Journal of analytical toxicology, 35(3), 183-187 (2011-03-29)
The Department of Defense (DoD) operates six forensic urine drug-testing laboratories that screen close to 5 million urine samples for amphetamines yearly. Recently, the DoD laboratories have observed a significant decrease in the confirmation rates for amphetamines because of specimens
Ju Hong Jeon et al.
FEBS letters, 534(1-3), 180-184 (2003-01-16)
Polyamine incorporation or cross-linking of proteins, post-translational modifications mediated by transglutaminase 2 (TGase 2), have been implicated in a variety of physiological functions including cell adhesion, extracellular matrix formation and apoptosis. To better understand the intracellular regulation mechanism of TGase
V V Novokhatny et al.
Thrombosis research, 53(3), 243-252 (1989-02-01)
The interaction of the isolated kringles 4 and 5 from human plasminogen with 6-aminohexanoic acid, pentylamine, pentanoic acid and arginine has been quantitatively characterized by scanning calorimetry and fluorescent spectroscopy. It has been found that the ligands with the positively

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service